Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46002973-46003218 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC61
Entrez Gene ID
729440
Gene Name
coiled-coil domain containing 61
Gene Aliases
-
UniGene
Hs.515479
Gene Chromosome Location
Chr.19: 45995457-46018616 on Build GRCh38
Transcripts
NM_001267723.1 XM_005259191.3 XM_005259192.4 XM_011527257.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3104921 c.277C>T p.R93W Chr.19: 46003124-46003124 on Build GRCh38
3104917 c.173T>C p.M58T Chr.19: 46003020-46003020 on Build GRCh38
998362 c.233G>A p.R78Q Chr.19: 46003080-46003080 on Build GRCh38
5028589 c.231G>A p.V77V Chr.19: 46003078-46003078 on Build GRCh38
6334536 c.278G>T p.R93L Chr.19: 46003125-46003125 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138444388 Chr.19: 46003029-46003029 on Build GRCh38
rs748214650 Chr.19: 46003145-46003145 on Build GRCh38
rs755617548 Chr.19: 46003010-46003010 on Build GRCh38
rs768269500 Chr.19: 46003176-46003176 on Build GRCh38
rs757804728 Chr.19: 46003100-46003100 on Build GRCh38
rs369923881 Chr.19: 46003030-46003030 on Build GRCh38
rs773301901 Chr.19: 46003074-46003074 on Build GRCh38
rs940260720 Chr.19: 46003187-46003187 on Build GRCh38
rs992393484 Chr.19: 46003121-46003121 on Build GRCh38
rs766527288 Chr.19: 46003003-46003003 on Build GRCh38
rs758619430 Chr.19: 46003117-46003117 on Build GRCh38
rs761303560 Chr.19: 46003179-46003179 on Build GRCh38
rs770957740 Chr.19: 46003151-46003151 on Build GRCh38
rs971207167 Chr.19: 46003063-46003063 on Build GRCh38
rs770958248 Chr.19: 46003076-46003076 on Build GRCh38
rs753477309 Chr.19: 46003054-46003054 on Build GRCh38
rs375888330 Chr.19: 46003075-46003075 on Build GRCh38
rs376202536 Chr.19: 46003115-46003115 on Build GRCh38
rs763548278 Chr.19: 46003093-46003093 on Build GRCh38
rs775232378 Chr.19: 46003170-46003170 on Build GRCh38
rs767054449 Chr.19: 46002998-46002998 on Build GRCh38
rs774249888 Chr.19: 46003078-46003078 on Build GRCh38
rs369405507 Chr.19: 46003124-46003124 on Build GRCh38
rs757643212 Chr.19: 46003045-46003045 on Build GRCh38
rs776170717 Chr.19: 46003178-46003178 on Build GRCh38
rs879449991 Chr.19: 46003024-46003024 on Build GRCh38
rs200638957 Chr.19: 46003165-46003165 on Build GRCh38
rs944140413 Chr.19: 46003161-46003161 on Build GRCh38
rs370094013 Chr.19: 46003058-46003058 on Build GRCh38
rs779298738 Chr.19: 46003046-46003046 on Build GRCh38
rs1017454491 Chr.19: 46003052-46003052 on Build GRCh38
rs371313075 Chr.19: 46003059-46003059 on Build GRCh38
rs372922369 Chr.19: 46003096-46003096 on Build GRCh38
rs149310015 Chr.19: 46003175-46003175 on Build GRCh38
rs753831019 Chr.19: 46003051-46003051 on Build GRCh38
rs754286303 Chr.19: 46003041-46003041 on Build GRCh38
rs887138581 Chr.19: 46003037-46003037 on Build GRCh38
rs1051089741 Chr.19: 46003034-46003034 on Build GRCh38
rs115054643 Chr.19: 46003132-46003132 on Build GRCh38
rs376628328 Chr.19: 46003089-46003089 on Build GRCh38
rs373145203 Chr.19: 46003125-46003125 on Build GRCh38
rs747113476 Chr.19: 46003065-46003065 on Build GRCh38
rs943214570 Chr.19: 46003028-46003028 on Build GRCh38
rs200651280 Chr.19: 46003156-46003156 on Build GRCh38
rs998705949 Chr.19: 46003180-46003180 on Build GRCh38
rs768794921 Chr.19: 46003066-46003066 on Build GRCh38
rs1049954005 Chr.19: 46003146-46003146 on Build GRCh38
rs933257309 Chr.19: 46003031-46003031 on Build GRCh38
rs759238493 Chr.19: 46003079-46003079 on Build GRCh38
rs760191941 Chr.19: 46003088-46003088 on Build GRCh38
rs369246676 Chr.19: 46003080-46003080 on Build GRCh38
rs527453041 Chr.19: 46003112-46003112 on Build GRCh38
rs761952377 Chr.19: 46003086-46003103 on Build GRCh38
rs201396254 Chr.19: 46003083-46003083 on Build GRCh38
rs377057292 Chr.19: 46003189-46003189 on Build GRCh38

View Full Product Details