Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 39266178-39266409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CX3CR1
Entrez Gene ID
1524
Gene Name
C-X3-C motif chemokine receptor 1
Gene Aliases
CCRL1, CMKBRL1, CMKDR1, GPR13, GPRV28, V28
UniGene
Hs.78913
Gene Chromosome Location
Chr.3: 39263494-39281735 on Build GRCh38
Transcripts
NM_001171171.1 NM_001171172.1 NM_001171174.1 NM_001337.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2987597 c.253C>T p.P85S Chr.3: 39266257-39266257 on Build GRCh38
1485167 c.295G>C p.D99H Chr.3: 39266311-39266311 on Build GRCh38
3728568 c.171C>T p.T57T Chr.3: 39266339-39266339 on Build GRCh38
3380574 c.161T>C p.F54S Chr.3: 39266349-39266349 on Build GRCh38
6097728 c.369G>A p.L123L Chr.3: 39266237-39266237 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767455598 Chr.3: 39266323-39266323 on Build GRCh38
rs548178853 Chr.3: 39266312-39266312 on Build GRCh38
rs780513445 Chr.3: 39266270-39266270 on Build GRCh38
rs769293817 Chr.3: 39266289-39266289 on Build GRCh38
rs367827486 Chr.3: 39266266-39266266 on Build GRCh38
rs561964760 Chr.3: 39266318-39266318 on Build GRCh38
rs753866039 Chr.3: 39266334-39266334 on Build GRCh38
rs754964383 Chr.3: 39266210-39266210 on Build GRCh38
rs938883415 Chr.3: 39266251-39266251 on Build GRCh38
rs753299596 Chr.3: 39266363-39266363 on Build GRCh38
rs776227760 Chr.3: 39266293-39266293 on Build GRCh38
rs780502032 Chr.3: 39266360-39266360 on Build GRCh38
rs758702644 Chr.3: 39266357-39266357 on Build GRCh38
rs779021239 Chr.3: 39266249-39266249 on Build GRCh38
rs747517244 Chr.3: 39266272-39266272 on Build GRCh38
rs750585901 Chr.3: 39266347-39266347 on Build GRCh38
rs372697125 Chr.3: 39266300-39266300 on Build GRCh38
rs751477915 Chr.3: 39266209-39266209 on Build GRCh38
rs766076052 Chr.3: 39266311-39266311 on Build GRCh38
rs748705144 Chr.3: 39266377-39266377 on Build GRCh38
rs746199750 Chr.3: 39266254-39266254 on Build GRCh38
rs191479599 Chr.3: 39266222-39266222 on Build GRCh38
rs781773493 Chr.3: 39266371-39266371 on Build GRCh38
rs757310143 Chr.3: 39266336-39266336 on Build GRCh38
rs546452868 Chr.3: 39266220-39266220 on Build GRCh38
rs199811198 Chr.3: 39266341-39266341 on Build GRCh38
rs759723364 Chr.3: 39266307-39266307 on Build GRCh38
rs761434711 Chr.3: 39266295-39266295 on Build GRCh38
rs531534991 Chr.3: 39266298-39266298 on Build GRCh38
rs777824021 Chr.3: 39266214-39266214 on Build GRCh38

View Full Product Details