Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 135910918-135911189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DARS
Entrez Gene ID
1615
Gene Name
aspartyl-tRNA synthetase
Gene Aliases
HBSL, aspRS
UniGene
Hs.503787
Gene Chromosome Location
Chr.2: 135905881-135986046 on Build GRCh38
Transcripts
NM_001293312.1 NM_001349.3 XM_017003489.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5803446 c.1396C>T p.H466Y Chr.2: 135911157-135911157 on Build GRCh38
5860137 c.1406G>A p.G469E Chr.2: 135911147-135911147 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs553265006 Chr.2: 135910997-135910997 on Build GRCh38
rs996466758 Chr.2: 135911031-135911031 on Build GRCh38
rs539747288 Chr.2: 135911097-135911097 on Build GRCh38
rs757088316 Chr.2: 135911120-135911120 on Build GRCh38
rs780897664 Chr.2: 135911165-135911165 on Build GRCh38
rs187084484 Chr.2: 135911051-135911051 on Build GRCh38
rs1042367232 Chr.2: 135910956-135910956 on Build GRCh38
rs899402326 Chr.2: 135911004-135911004 on Build GRCh38
rs780386560 Chr.2: 135911090-135911090 on Build GRCh38
rs557510868 Chr.2: 135911127-135911127 on Build GRCh38
rs753644168 Chr.2: 135911106-135911106 on Build GRCh38
rs1021995580 Chr.2: 135911163-135911163 on Build GRCh38
rs757872233 Chr.2: 135911144-135911144 on Build GRCh38
rs746657526 Chr.2: 135911156-135911156 on Build GRCh38
rs940705522 Chr.2: 135910964-135910964 on Build GRCh38
rs944017201 Chr.2: 135910953-135910953 on Build GRCh38
rs1037731246 Chr.2: 135910983-135910983 on Build GRCh38
rs148806569 Chr.2: 135911162-135911162 on Build GRCh38
rs774685673 Chr.2: 135911099-135911099 on Build GRCh38
rs763936828 Chr.2: 135911100-135911100 on Build GRCh38
rs779584968 Chr.2: 135911152-135911152 on Build GRCh38
rs539573634 Chr.2: 135910951-135910951 on Build GRCh38
rs766145716 Chr.2: 135911093-135911093 on Build GRCh38
rs892237452 Chr.2: 135911126-135911126 on Build GRCh38
rs750390582 Chr.2: 135911131-135911131 on Build GRCh38
rs573403535 Chr.2: 135911050-135911050 on Build GRCh38
rs115253602 Chr.2: 135911121-135911121 on Build GRCh38
rs751745838 Chr.2: 135911154-135911154 on Build GRCh38

View Full Product Details