Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 62474166-62474393 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CADPS
Entrez Gene ID
8618
Gene Name
calcium dependent secretion activator
Gene Aliases
CADPS1, CAPS, CAPS1, UNC-31
UniGene
Hs.654933
Gene Chromosome Location
Chr.3: 62398346-62875389 on Build GRCh38
Transcripts
NM_003716.3 NM_183393.2 NM_183394.2 XM_006713378.2 XM_011534177.1 XM_011534178.1 XM_011534180.1 XM_011534182.1 XM_011534183.1 XM_011534189.1 XM_011534191.2 XM_011534192.1 XM_011534193.1 XM_011534194.2 XM_011534195.1 XM_011534196.1 XM_011534197.2 XM_011534198.2 XM_011534199.2 XM_011534200.2 XM_011534203.1 XM_017007358.1 XM_017007359.1 XM_017007360.1 XM_017007361.1 XM_017007362.1 XM_017007363.1 XM_017007364.1 XM_017007365.1 XM_017007366.1 XM_017007367.1 XM_017007368.1 XM_017007369.1 XM_017007370.1 XM_017007371.1 XM_017007372.1 XM_017007373.1 XM_017007374.1 XM_017007375.1 XM_017007376.1 XM_017007377.1 XM_017007378.1 XM_017007379.1 XM_017007380.1 XM_017007381.1 XM_017007382.1 XM_017007383.1 XM_017007384.1 XM_017007385.1 XM_017007386.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2783097 c.3404G>A p.C1135Y Chr.3: 62474246-62474246 on Build GRCh38
6618880 c.3383T>G p.F1128C Chr.3: 62474267-62474267 on Build GRCh38
1692939 c.3385C>T p.R1129* Chr.3: 62474265-62474265 on Build GRCh38
1047930 c.3273C>T p.V1091V Chr.3: 62474260-62474260 on Build GRCh38
1425025 c.3274C>T p.P1092S Chr.3: 62474259-62474259 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753913796 Chr.3: 62474257-62474257 on Build GRCh38
rs776749191 Chr.3: 62474209-62474209 on Build GRCh38
rs920892751 Chr.3: 62474269-62474269 on Build GRCh38
rs149931152 Chr.3: 62474312-62474312 on Build GRCh38
rs760213417 Chr.3: 62474362-62474362 on Build GRCh38
rs1029699167 Chr.3: 62474363-62474363 on Build GRCh38
rs761716002 Chr.3: 62474354-62474354 on Build GRCh38
rs148854672 Chr.3: 62474278-62474278 on Build GRCh38
rs763771626 Chr.3: 62474226-62474226 on Build GRCh38
rs151301388 Chr.3: 62474285-62474285 on Build GRCh38
rs201181624 Chr.3: 62474280-62474280 on Build GRCh38
rs961507723 Chr.3: 62474348-62474348 on Build GRCh38
rs778256058 Chr.3: 62474279-62474279 on Build GRCh38
rs781377421 Chr.3: 62474310-62474310 on Build GRCh38
rs112585082 Chr.3: 62474236-62474236 on Build GRCh38
rs771021661 Chr.3: 62474298-62474298 on Build GRCh38
rs369528205 Chr.3: 62474368-62474368 on Build GRCh38
rs753456107 Chr.3: 62474255-62474255 on Build GRCh38
rs766019189 Chr.3: 62474253-62474253 on Build GRCh38
rs748581222 Chr.3: 62474265-62474265 on Build GRCh38
rs372021165 Chr.3: 62474355-62474355 on Build GRCh38
rs754709322 Chr.3: 62474260-62474260 on Build GRCh38
rs774239450 Chr.3: 62474230-62474230 on Build GRCh38
rs534391982 Chr.3: 62474237-62474237 on Build GRCh38
rs367714345 Chr.3: 62474264-62474264 on Build GRCh38
rs774001539 Chr.3: 62474337-62474337 on Build GRCh38
rs199647496 Chr.3: 62474250-62474250 on Build GRCh38
rs761834820 Chr.3: 62474232-62474232 on Build GRCh38
rs762756786 Chr.3: 62474224-62474224 on Build GRCh38

View Full Product Details