Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 44289235-44289460 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASC4
Entrez Gene ID
113201
Gene Name
cancer susceptibility candidate 4
Gene Aliases
H63
UniGene
Hs.512867
Gene Chromosome Location
Chr.15: 44288616-44415761 on Build GRCh38
Transcripts
NM_138423.3 NM_177974.2 XM_017021880.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7400445 c.228G>A p.T76T Chr.15: 44289257-44289257 on Build GRCh38
1749166 c.273C>T p.L91L Chr.15: 44289302-44289302 on Build GRCh38
3386887 c.279C>A p.S93R Chr.15: 44289308-44289308 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764537544 Chr.15: 44289407-44289407 on Build GRCh38
rs59997416 Chr.15: 44289402-44289402 on Build GRCh38
rs910301500 Chr.15: 44289409-44289409 on Build GRCh38
rs747082945 Chr.15: 44289322-44289322 on Build GRCh38
rs766990879 Chr.15: 44289278-44289278 on Build GRCh38
rs778318372 Chr.15: 44289289-44289289 on Build GRCh38
rs371714392 Chr.15: 44289290-44289290 on Build GRCh38
rs752242710 Chr.15: 44289282-44289282 on Build GRCh38
rs752194987 Chr.15: 44289392-44289392 on Build GRCh38
rs769669887 Chr.15: 44289333-44289333 on Build GRCh38
rs746031745 Chr.15: 44289309-44289309 on Build GRCh38
rs772921889 Chr.15: 44289255-44289255 on Build GRCh38
rs763287981 Chr.15: 44289340-44289340 on Build GRCh38
rs944423395 Chr.15: 44289421-44289421 on Build GRCh38
rs992244243 Chr.15: 44289430-44289430 on Build GRCh38
rs576965613 Chr.15: 44289319-44289319 on Build GRCh38
rs770786408 Chr.15: 44289348-44289348 on Build GRCh38
rs767164436 Chr.15: 44289389-44289389 on Build GRCh38
rs200133088 Chr.15: 44289277-44289277 on Build GRCh38
rs762822512 Chr.15: 44289256-44289256 on Build GRCh38
rs901991379 Chr.15: 44289267-44289267 on Build GRCh38
rs1040027337 Chr.15: 44289426-44289426 on Build GRCh38
rs759325430 Chr.15: 44289374-44289374 on Build GRCh38
rs142240634 Chr.15: 44289310-44289310 on Build GRCh38
rs148489550 Chr.15: 44289263-44289263 on Build GRCh38
rs756617189 Chr.15: 44289288-44289288 on Build GRCh38
rs540145112 Chr.15: 44289294-44289294 on Build GRCh38
rs779112352 Chr.15: 44289304-44289304 on Build GRCh38
rs545950464 Chr.15: 44289386-44289386 on Build GRCh38
rs751221524 Chr.15: 44289274-44289274 on Build GRCh38
rs774270905 Chr.15: 44289356-44289356 on Build GRCh38
rs962074535 Chr.15: 44289415-44289415 on Build GRCh38
rs762874987 Chr.15: 44289335-44289335 on Build GRCh38

View Full Product Details