Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 37256317-37256584 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GHRH
Entrez Gene ID
2691
Gene Name
growth hormone releasing hormone
Gene Aliases
GHRF, GRF, INN
UniGene
Hs.37023
Gene Chromosome Location
Chr.20: 37251086-37261817 on Build GRCh38
Transcripts
NM_001184731.2 NM_021081.5 XM_011528784.2 XM_011528788.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6046910 c.84-6G>A p.? Chr.20: 37256504-37256504 on Build GRCh38
1026505 c.88C>T p.R30W Chr.20: 37256494-37256494 on Build GRCh38
4098142 c.119G>A p.S40N Chr.20: 37256463-37256463 on Build GRCh38
123354 c.84G>T p.R28S Chr.20: 37256498-37256498 on Build GRCh38
3546046 c.171C>T p.I57I Chr.20: 37256411-37256411 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770886854 Chr.20: 37256417-37256417 on Build GRCh38
rs775688947 Chr.20: 37256519-37256519 on Build GRCh38
rs779536089 Chr.20: 37256356-37256356 on Build GRCh38
rs550735569 Chr.20: 37256562-37256562 on Build GRCh38
rs770654030 Chr.20: 37256490-37256490 on Build GRCh38
rs367766804 Chr.20: 37256528-37256528 on Build GRCh38
rs760643047 Chr.20: 37256524-37256524 on Build GRCh38
rs758823653 Chr.20: 37256463-37256463 on Build GRCh38
rs371047125 Chr.20: 37256440-37256440 on Build GRCh38
rs769420800 Chr.20: 37256397-37256397 on Build GRCh38
rs150553254 Chr.20: 37256435-37256435 on Build GRCh38
rs371321167 Chr.20: 37256402-37256402 on Build GRCh38
rs893049345 Chr.20: 37256557-37256557 on Build GRCh38
rs28932185 Chr.20: 37256476-37256476 on Build GRCh38
rs749103484 Chr.20: 37256489-37256489 on Build GRCh38
rs777362452 Chr.20: 37256484-37256484 on Build GRCh38
rs780809718 Chr.20: 37256468-37256468 on Build GRCh38
rs772701000 Chr.20: 37256408-37256408 on Build GRCh38
rs755803548 Chr.20: 37256480-37256480 on Build GRCh38
rs990835110 Chr.20: 37256457-37256457 on Build GRCh38
rs368475481 Chr.20: 37256430-37256430 on Build GRCh38
rs566092278 Chr.20: 37256434-37256434 on Build GRCh38
rs376701954 Chr.20: 37256358-37256358 on Build GRCh38
rs371555766 Chr.20: 37256529-37256529 on Build GRCh38
rs17787698 Chr.20: 37256487-37256487 on Build GRCh38
rs754464713 Chr.20: 37256363-37256363 on Build GRCh38
rs765760128 Chr.20: 37256545-37256545 on Build GRCh38
rs765871814 Chr.20: 37256458-37256458 on Build GRCh38
rs535603134 Chr.20: 37256530-37256530 on Build GRCh38
rs970111387 Chr.20: 37256438-37256438 on Build GRCh38
rs749049523 Chr.20: 37256413-37256413 on Build GRCh38
rs759549389 Chr.20: 37256431-37256431 on Build GRCh38
rs1015216356 Chr.20: 37256406-37256406 on Build GRCh38
rs981258896 Chr.20: 37256496-37256496 on Build GRCh38
rs6032441 Chr.20: 37256516-37256516 on Build GRCh38
rs989077559 Chr.20: 37256565-37256565 on Build GRCh38
rs754297738 Chr.20: 37256447-37256447 on Build GRCh38
rs762233133 Chr.20: 37256453-37256453 on Build GRCh38
rs552944131 Chr.20: 37256377-37256377 on Build GRCh38
rs750872486 Chr.20: 37256460-37256460 on Build GRCh38
rs373627645 Chr.20: 37256504-37256504 on Build GRCh38
rs374317089 Chr.20: 37256505-37256505 on Build GRCh38
rs776215379 Chr.20: 37256554-37256554 on Build GRCh38
rs747897519 Chr.20: 37256384-37256384 on Build GRCh38
rs778720800 Chr.20: 37256493-37256493 on Build GRCh38

View Full Product Details