Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 61743554-61743827 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH4
Entrez Gene ID
1002
Gene Name
cadherin 4
Gene Aliases
CAD4, R-CAD, RCAD
UniGene
Hs.473231
Gene Chromosome Location
Chr.20: 61252426-61940617 on Build GRCh38
Transcripts
NM_001252338.2 NM_001252339.2 NM_001794.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6558957 c.198G>T p.K66N Chr.20: 61743591-61743591 on Build GRCh38
5513643 c.342C>T p.D114D Chr.20: 61743735-61743735 on Build GRCh38
724920 c.391C>A p.H131N Chr.20: 61743784-61743784 on Build GRCh38
4963169 c.352C>T p.R118W Chr.20: 61743745-61743745 on Build GRCh38
4409086 c.231C>A p.F77L Chr.20: 61743624-61743624 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764246663 Chr.20: 61743802-61743802 on Build GRCh38
rs760527005 Chr.20: 61743739-61743739 on Build GRCh38
rs772958577 Chr.20: 61743736-61743736 on Build GRCh38
rs369039825 Chr.20: 61743745-61743745 on Build GRCh38
rs375789487 Chr.20: 61743699-61743699 on Build GRCh38
rs760401271 Chr.20: 61743630-61743630 on Build GRCh38
rs1033951496 Chr.20: 61743711-61743711 on Build GRCh38
rs79132902 Chr.20: 61743690-61743690 on Build GRCh38
rs569045230 Chr.20: 61743696-61743696 on Build GRCh38
rs147587513 Chr.20: 61743774-61743774 on Build GRCh38
rs752749815 Chr.20: 61743741-61743741 on Build GRCh38
rs761709044 Chr.20: 61743659-61743659 on Build GRCh38
rs142221912 Chr.20: 61743651-61743651 on Build GRCh38
rs145264164 Chr.20: 61743729-61743729 on Build GRCh38
rs778014823 Chr.20: 61743693-61743693 on Build GRCh38
rs776377387 Chr.20: 61743735-61743735 on Build GRCh38
rs182629710 Chr.20: 61743599-61743599 on Build GRCh38
rs779698363 Chr.20: 61743709-61743709 on Build GRCh38
rs548726503 Chr.20: 61743594-61743594 on Build GRCh38
rs754111063 Chr.20: 61743759-61743759 on Build GRCh38
rs773043144 Chr.20: 61743786-61743786 on Build GRCh38
rs769403401 Chr.20: 61743779-61743779 on Build GRCh38
rs1040086720 Chr.20: 61743657-61743657 on Build GRCh38
rs975368737 Chr.20: 61743748-61743748 on Build GRCh38
rs765048505 Chr.20: 61743664-61743664 on Build GRCh38
rs750401864 Chr.20: 61743670-61743670 on Build GRCh38
rs371518012 Chr.20: 61743768-61743768 on Build GRCh38
rs147816904 Chr.20: 61743672-61743672 on Build GRCh38
rs139681917 Chr.20: 61743713-61743713 on Build GRCh38
rs767592477 Chr.20: 61743678-61743678 on Build GRCh38
rs368305378 Chr.20: 61743724-61743724 on Build GRCh38
rs187771644 Chr.20: 61743806-61743806 on Build GRCh38
rs375223593 Chr.20: 61743807-61743807 on Build GRCh38
rs771640620 Chr.20: 61743705-61743705 on Build GRCh38
rs753938834 Chr.20: 61743803-61743803 on Build GRCh38
rs762783096 Chr.20: 61743787-61743787 on Build GRCh38
rs745328834 Chr.20: 61743614-61743614 on Build GRCh38
rs888923510 Chr.20: 61743635-61743635 on Build GRCh38
rs780747185 Chr.20: 61743770-61743770 on Build GRCh38
rs373320390 Chr.20: 61743746-61743746 on Build GRCh38
rs768392719 Chr.20: 61743721-61743721 on Build GRCh38
rs367543860 Chr.20: 61743801-61743801 on Build GRCh38
rs993300563 Chr.20: 61743681-61743681 on Build GRCh38
rs762875694 Chr.20: 61743738-61743738 on Build GRCh38
rs749580573 Chr.20: 61743695-61743695 on Build GRCh38
rs776268387 Chr.20: 61743652-61743652 on Build GRCh38
rs61737806 Chr.20: 61743767-61743767 on Build GRCh38
rs1005061662 Chr.20: 61743589-61743589 on Build GRCh38
rs375901541 Chr.20: 61743794-61743794 on Build GRCh38
rs746478957 Chr.20: 61743771-61743771 on Build GRCh38
rs757506706 Chr.20: 61743761-61743761 on Build GRCh38
rs771730396 Chr.20: 61743617-61743617 on Build GRCh38
rs775223229 Chr.20: 61743618-61743618 on Build GRCh38
rs774300371 Chr.20: 61743795-61743795 on Build GRCh38
rs746397917 Chr.20: 61743704-61743704 on Build GRCh38
rs752838009 Chr.20: 61743689-61743689 on Build GRCh38
rs780655670 Chr.20: 61743773-61743773 on Build GRCh38

View Full Product Details