Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7581323-7581586 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD68
Entrez Gene ID
968
Gene Name
CD68 molecule
Gene Aliases
GP110, LAMP4, SCARD1
UniGene
Hs.647419
Gene Chromosome Location
Chr.17: 7579487-7582111 on Build GRCh38
Transcripts
NM_001040059.1 NM_001251.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2743869 c.932-6C>T p.? Chr.17: 7581372-7581372 on Build GRCh38
3766187 c.1048G>A p.A350T Chr.17: 7581494-7581494 on Build GRCh38
6249655 c.954G>T p.R318R Chr.17: 7581400-7581400 on Build GRCh38
1709722 c.1043C>T p.P348L Chr.17: 7581489-7581489 on Build GRCh38
6222213 c.1034G>A p.R345Q Chr.17: 7581480-7581480 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755041269 Chr.17: 7581480-7581480 on Build GRCh38
rs368188387 Chr.17: 7581538-7581538 on Build GRCh38
rs773716363 Chr.17: 7581405-7581405 on Build GRCh38
rs991768651 Chr.17: 7581424-7581424 on Build GRCh38
rs963720197 Chr.17: 7581560-7581561 on Build GRCh38
rs749196214 Chr.17: 7581363-7581363 on Build GRCh38
rs770108113 Chr.17: 7581402-7581402 on Build GRCh38
rs781304672 Chr.17: 7581482-7581482 on Build GRCh38
rs770159428 Chr.17: 7581485-7581485 on Build GRCh38
rs377723278 Chr.17: 7581395-7581395 on Build GRCh38
rs951312054 Chr.17: 7581372-7581373 on Build GRCh38
rs761679144 Chr.17: 7581537-7581537 on Build GRCh38
rs780340661 Chr.17: 7581372-7581372 on Build GRCh38
rs751096463 Chr.17: 7581547-7581547 on Build GRCh38
rs942526546 Chr.17: 7581533-7581533 on Build GRCh38
rs755915279 Chr.17: 7581357-7581357 on Build GRCh38
rs763835276 Chr.17: 7581523-7581523 on Build GRCh38
rs758554225 Chr.17: 7581368-7581368 on Build GRCh38
rs752344735 Chr.17: 7581425-7581425 on Build GRCh38
rs759209711 Chr.17: 7581557-7581557 on Build GRCh38
rs751640523 Chr.17: 7581479-7581479 on Build GRCh38
rs372695114 Chr.17: 7581373-7581373 on Build GRCh38
rs187070664 Chr.17: 7581534-7581534 on Build GRCh38
rs370867284 Chr.17: 7581492-7581492 on Build GRCh38
rs761553819 Chr.17: 7581416-7581416 on Build GRCh38
rs771570633 Chr.17: 7581511-7581511 on Build GRCh38
rs35452170 Chr.17: 7581432-7581432 on Build GRCh38
rs140596584 Chr.17: 7581472-7581472 on Build GRCh38
rs374543382 Chr.17: 7581520-7581520 on Build GRCh38
rs763999884 Chr.17: 7581428-7581428 on Build GRCh38
rs752471089 Chr.17: 7581350-7581350 on Build GRCh38
rs142443844 Chr.17: 7581512-7581512 on Build GRCh38
rs763912874 Chr.17: 7581439-7581439 on Build GRCh38
rs17607 Chr.17: 7581464-7581464 on Build GRCh38
rs922374101 Chr.17: 7581434-7581434 on Build GRCh38
rs150041207 Chr.17: 7581493-7581493 on Build GRCh38
rs777874530 Chr.17: 7581358-7581358 on Build GRCh38
rs34688956 Chr.17: 7581563-7581564 on Build GRCh38
rs758990817 Chr.17: 7581495-7581495 on Build GRCh38
rs767124041 Chr.17: 7581422-7581422 on Build GRCh38
rs188221678 Chr.17: 7581419-7581419 on Build GRCh38
rs9901675 Chr.17: 7581494-7581494 on Build GRCh38
rs755021833 Chr.17: 7581542-7581542 on Build GRCh38
rs1056303677 Chr.17: 7581496-7581496 on Build GRCh38
rs374100573 Chr.17: 7581440-7581440 on Build GRCh38
rs933509671 Chr.17: 7581448-7581448 on Build GRCh38
rs946301105 Chr.17: 7581486-7581486 on Build GRCh38
rs142060051 Chr.17: 7581443-7581443 on Build GRCh38
rs145691004 Chr.17: 7581540-7581540 on Build GRCh38
rs750900821 Chr.17: 7581562-7581563 on Build GRCh38
rs748487888 Chr.17: 7581399-7581399 on Build GRCh38
rs981540329 Chr.17: 7581389-7581389 on Build GRCh38
rs778166210 Chr.17: 7581487-7581487 on Build GRCh38
rs763395779 Chr.17: 7581410-7581410 on Build GRCh38
rs200380131 Chr.17: 7581398-7581398 on Build GRCh38
rs545356456 Chr.17: 7581556-7581557 on Build GRCh38
rs540120096 Chr.17: 7581345-7581345 on Build GRCh38
rs781278034 Chr.17: 7581545-7581545 on Build GRCh38

View Full Product Details