Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27100359-27100632 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CGREF1
Entrez Gene ID
10669
Gene Name
cell growth regulator with EF-hand domain 1
Gene Aliases
CGR11
UniGene
Hs.159525
Gene Chromosome Location
Chr.2: 27099353-27119127 on Build GRCh38
Transcripts
NM_001166239.1 NM_001166240.1 NM_001301324.1 NM_006569.5 NM_000221.2 NM_006488.2 XM_005264294.3 XM_005264296.3 XM_005264298.3 XM_017004060.1 XM_017004061.1

Gene Symbol
KHK
Entrez Gene ID
3795
Gene Name
ketohexokinase
Gene Aliases
-
UniGene
Hs.567297
Gene Chromosome Location
Chr.2: 27086738-27100751 on Build GRCh38
Transcripts
NM_001166239.1 NM_001166240.1 NM_001301324.1 NM_006569.5 NM_000221.2 NM_006488.2 XM_005264294.3 XM_005264296.3 XM_005264298.3 XM_017004060.1 XM_017004061.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6349972 c.181G>A p.G61S Chr.2: 27100517-27100517 on Build GRCh38
1631949 c.171T>C p.Y57Y Chr.2: 27100527-27100527 on Build GRCh38
5726203 c.192C>T p.S64S Chr.2: 27100506-27100506 on Build GRCh38
1019520 c.158C>A p.P53H Chr.2: 27100540-27100540 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs576618906 Chr.2: 27100559-27100559 on Build GRCh38
rs189380151 Chr.2: 27100572-27100572 on Build GRCh38
rs779916668 Chr.2: 27100460-27100460 on Build GRCh38
rs777666196 Chr.2: 27100540-27100540 on Build GRCh38
rs962315515 Chr.2: 27100410-27100410 on Build GRCh38
rs990611110 Chr.2: 27100515-27100515 on Build GRCh38
rs1007451301 Chr.2: 27100593-27100593 on Build GRCh38
rs886055894 Chr.2: 27100541-27100541 on Build GRCh38
rs769822651 Chr.2: 27100550-27100550 on Build GRCh38
rs536842595 Chr.2: 27100382-27100382 on Build GRCh38
rs973719326 Chr.2: 27100414-27100414 on Build GRCh38
rs769571556 Chr.2: 27100526-27100526 on Build GRCh38
rs17847233 Chr.2: 27100502-27100502 on Build GRCh38
rs976707941 Chr.2: 27100465-27100465 on Build GRCh38
rs543665606 Chr.2: 27100560-27100560 on Build GRCh38
rs965373017 Chr.2: 27100392-27100392 on Build GRCh38
rs758800413 Chr.2: 27100455-27100455 on Build GRCh38
rs766710172 Chr.2: 27100501-27100502 on Build GRCh38
rs936498561 Chr.2: 27100506-27100506 on Build GRCh38
rs750863687 Chr.2: 27100449-27100449 on Build GRCh38
rs1046075237 Chr.2: 27100552-27100552 on Build GRCh38
rs888658723 Chr.2: 27100588-27100588 on Build GRCh38
rs570660611 Chr.2: 27100403-27100403 on Build GRCh38
rs1049111395 Chr.2: 27100583-27100583 on Build GRCh38
rs73921452 Chr.2: 27100504-27100504 on Build GRCh38
rs184162960 Chr.2: 27100401-27100401 on Build GRCh38
rs4665371 Chr.2: 27100600-27100600 on Build GRCh38
rs751242318 Chr.2: 27100489-27100489 on Build GRCh38
rs141247751 Chr.2: 27100463-27100463 on Build GRCh38
rs113859516 Chr.2: 27100556-27100556 on Build GRCh38
rs992176466 Chr.2: 27100501-27100501 on Build GRCh38
rs1030217877 Chr.2: 27100486-27100486 on Build GRCh38
rs1131375 Chr.2: 27100517-27100517 on Build GRCh38
rs576893423 Chr.2: 27100500-27100500 on Build GRCh38

View Full Product Details