Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 109769108-109769361 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM222A
Entrez Gene ID
84915
Gene Name
family with sequence similarity 222 member A
Gene Aliases
C12orf34
UniGene
Hs.661785
Gene Chromosome Location
Chr.12: 109714382-109770509 on Build GRCh38
Transcripts
NM_032829.2 XM_006719654.2 XM_017020055.1 NR_026661.2 NR_026662.2

Gene Symbol
FAM222A-AS1
Entrez Gene ID
84983
Gene Name
FAM222A antisense RNA 1
Gene Aliases
-
UniGene
Hs.675961
Gene Chromosome Location
Chr.12: 109734209-109773487 on Build GRCh38
Transcripts
NM_032829.2 XM_006719654.2 XM_017020055.1 NR_026661.2 NR_026662.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7279844 c.1274T>C p.M425T Chr.12: 109769203-109769203 on Build GRCh38
4038585 c.1190G>A p.C397Y Chr.12: 109769119-109769119 on Build GRCh38
5507494 c.1350C>G p.V450V Chr.12: 109769279-109769279 on Build GRCh38
7176982 c.1347C>T p.P449P Chr.12: 109769276-109769276 on Build GRCh38
115783 c.1182G>C p.K394N Chr.12: 109769111-109769111 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1009016158 Chr.12: 109769257-109769257 on Build GRCh38
rs757498539 Chr.12: 109769308-109769315 on Build GRCh38
rs554087178 Chr.12: 109769295-109769295 on Build GRCh38
rs578010640 Chr.12: 109769274-109769274 on Build GRCh38
rs750523692 Chr.12: 109769301-109769301 on Build GRCh38
rs150508057 Chr.12: 109769273-109769273 on Build GRCh38
rs781555563 Chr.12: 109769315-109769315 on Build GRCh38
rs538907659 Chr.12: 109769292-109769292 on Build GRCh38
rs372263353 Chr.12: 109769317-109769317 on Build GRCh38
rs758976307 Chr.12: 109769236-109769236 on Build GRCh38
rs375066166 Chr.12: 109769325-109769325 on Build GRCh38
rs758353875 Chr.12: 109769306-109769306 on Build GRCh38
rs193020897 Chr.12: 109769300-109769300 on Build GRCh38
rs769385983 Chr.12: 109769265-109769265 on Build GRCh38
rs755452651 Chr.12: 109769313-109769313 on Build GRCh38
rs746428249 Chr.12: 109769143-109769143 on Build GRCh38
rs867798744 Chr.12: 109769260-109769260 on Build GRCh38
rs747299998 Chr.12: 109769163-109769163 on Build GRCh38
rs1034634299 Chr.12: 109769271-109769271 on Build GRCh38
rs142286258 Chr.12: 109769241-109769241 on Build GRCh38
rs769330810 Chr.12: 109769172-109769172 on Build GRCh38
rs762149616 Chr.12: 109769297-109769297 on Build GRCh38
rs200835101 Chr.12: 109769179-109769179 on Build GRCh38
rs772086317 Chr.12: 109769334-109769334 on Build GRCh38
rs962002418 Chr.12: 109769307-109769307 on Build GRCh38
rs772438317 Chr.12: 109769144-109769144 on Build GRCh38
rs753250796 Chr.12: 109769297-109769299 on Build GRCh38
rs757523116 Chr.12: 109769231-109769231 on Build GRCh38
rs780361445 Chr.12: 109769238-109769238 on Build GRCh38
rs779790772 Chr.12: 109769325-109769344 on Build GRCh38
rs762302081 Chr.12: 109769269-109769269 on Build GRCh38
rs971243805 Chr.12: 109769309-109769309 on Build GRCh38
rs752076569 Chr.12: 109769308-109769308 on Build GRCh38
rs778170329 Chr.12: 109769299-109769300 on Build GRCh38
rs184096985 Chr.12: 109769224-109769224 on Build GRCh38
rs368611481 Chr.12: 109769277-109769277 on Build GRCh38
rs776212609 Chr.12: 109769170-109769170 on Build GRCh38
rs754243612 Chr.12: 109769300-109769307 on Build GRCh38
rs201840129 Chr.12: 109769239-109769239 on Build GRCh38
rs934573779 Chr.12: 109769345-109769345 on Build GRCh38
rs781502350 Chr.12: 109769246-109769246 on Build GRCh38
rs767460451 Chr.12: 109769290-109769290 on Build GRCh38
rs769751906 Chr.12: 109769297-109769298 on Build GRCh38
rs745850582 Chr.12: 109769326-109769326 on Build GRCh38
rs149644511 Chr.12: 109769183-109769183 on Build GRCh38
rs751372086 Chr.12: 109769204-109769204 on Build GRCh38
rs761446019 Chr.12: 109769171-109769171 on Build GRCh38
rs776580547 Chr.12: 109769232-109769232 on Build GRCh38
rs532625584 Chr.12: 109769166-109769166 on Build GRCh38
rs754179312 Chr.12: 109769228-109769228 on Build GRCh38
rs774213903 Chr.12: 109769276-109769276 on Build GRCh38
rs780490812 Chr.12: 109769153-109769153 on Build GRCh38
rs537650643 Chr.12: 109769268-109769268 on Build GRCh38
rs138993259 Chr.12: 109769225-109769225 on Build GRCh38
rs748470704 Chr.12: 109769316-109769316 on Build GRCh38
rs892898069 Chr.12: 109769192-109769192 on Build GRCh38
rs747843008 Chr.12: 109769247-109769247 on Build GRCh38
rs766184124 Chr.12: 109769196-109769196 on Build GRCh38
rs907142663 Chr.12: 109769258-109769258 on Build GRCh38
rs560864515 Chr.12: 109769340-109769340 on Build GRCh38
rs779131434 Chr.12: 109769129-109769129 on Build GRCh38

View Full Product Details