Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96828488-96828753 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNNM3
Entrez Gene ID
26505
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 3
Gene Aliases
ACDP3
UniGene
Hs.711617
Gene Chromosome Location
Chr.2: 96815894-96837109 on Build GRCh38
Transcripts
NM_017623.4 NM_199078.2 XM_011510957.2 XM_017003800.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3391901 c.1799C>T p.P600L Chr.2: 96828579-96828579 on Build GRCh38
5491421 c.1822C>T p.R608C Chr.2: 96828602-96828602 on Build GRCh38
2825840 c.1846G>A p.G616S Chr.2: 96828626-96828626 on Build GRCh38
4673150 c.1876C>T p.P626S Chr.2: 96828656-96828656 on Build GRCh38
160070 c.1857G>A p.T619T Chr.2: 96828637-96828637 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200074384 Chr.2: 96828730-96828730 on Build GRCh38
rs577439389 Chr.2: 96828619-96828619 on Build GRCh38
rs747437794 Chr.2: 96828719-96828719 on Build GRCh38
rs529324762 Chr.2: 96828667-96828667 on Build GRCh38
rs372047605 Chr.2: 96828649-96828649 on Build GRCh38
rs143099366 Chr.2: 96828563-96828563 on Build GRCh38
rs774682167 Chr.2: 96828580-96828580 on Build GRCh38
rs772032444 Chr.2: 96828554-96828554 on Build GRCh38
rs1020255800 Chr.2: 96828589-96828589 on Build GRCh38
rs774186802 Chr.2: 96828631-96828631 on Build GRCh38
rs775042700 Chr.2: 96828646-96828646 on Build GRCh38
rs773447518 Chr.2: 96828707-96828707 on Build GRCh38
rs755577067 Chr.2: 96828684-96828684 on Build GRCh38
rs757171251 Chr.2: 96828680-96828680 on Build GRCh38
rs926796639 Chr.2: 96828525-96828525 on Build GRCh38
rs148219000 Chr.2: 96828648-96828648 on Build GRCh38
rs201906694 Chr.2: 96828683-96828683 on Build GRCh38
rs781153668 Chr.2: 96828681-96828681 on Build GRCh38
rs768033531 Chr.2: 96828590-96828590 on Build GRCh38
rs762130482 Chr.2: 96828588-96828588 on Build GRCh38
rs769410936 Chr.2: 96828637-96828637 on Build GRCh38
rs779642440 Chr.2: 96828686-96828686 on Build GRCh38
rs766512708 Chr.2: 96828597-96828597 on Build GRCh38
rs1003317834 Chr.2: 96828599-96828599 on Build GRCh38
rs777645164 Chr.2: 96828620-96828620 on Build GRCh38
rs778781046 Chr.2: 96828545-96828545 on Build GRCh38
rs146508149 Chr.2: 96828659-96828659 on Build GRCh38
rs374033972 Chr.2: 96828658-96828658 on Build GRCh38
rs750859050 Chr.2: 96828592-96828592 on Build GRCh38
rs754060852 Chr.2: 96828538-96828538 on Build GRCh38
rs148563074 Chr.2: 96828636-96828636 on Build GRCh38
rs780307848 Chr.2: 96828534-96828534 on Build GRCh38
rs945435453 Chr.2: 96828575-96828575 on Build GRCh38
rs751437154 Chr.2: 96828675-96828675 on Build GRCh38
rs771544489 Chr.2: 96828724-96828724 on Build GRCh38
rs752534340 Chr.2: 96828676-96828676 on Build GRCh38
rs750497289 Chr.2: 96828531-96828531 on Build GRCh38
rs149864695 Chr.2: 96828603-96828603 on Build GRCh38
rs749129897 Chr.2: 96828518-96828519 on Build GRCh38
rs759120670 Chr.2: 96828578-96828578 on Build GRCh38
rs756192254 Chr.2: 96828532-96828532 on Build GRCh38
rs796066509 Chr.2: 96828701-96828701 on Build GRCh38
rs773642498 Chr.2: 96828651-96828651 on Build GRCh38
rs770379980 Chr.2: 96828570-96828570 on Build GRCh38
rs943741502 Chr.2: 96828508-96828508 on Build GRCh38
rs770701630 Chr.2: 96828610-96828610 on Build GRCh38
rs778499301 Chr.2: 96828557-96828557 on Build GRCh38
rs759739621 Chr.2: 96828665-96828665 on Build GRCh38
rs748837524 Chr.2: 96828691-96828691 on Build GRCh38
rs776838342 Chr.2: 96828662-96828662 on Build GRCh38
rs780674980 Chr.2: 96828632-96828632 on Build GRCh38
rs199790791 Chr.2: 96828602-96828602 on Build GRCh38
rs765526017 Chr.2: 96828666-96828666 on Build GRCh38
rs529615631 Chr.2: 96828709-96828709 on Build GRCh38
rs1027747782 Chr.2: 96828694-96828694 on Build GRCh38
rs769947090 Chr.2: 96828571-96828571 on Build GRCh38
rs746497730 Chr.2: 96828569-96828569 on Build GRCh38
rs754286149 Chr.2: 96828668-96828668 on Build GRCh38
rs768362653 Chr.2: 96828700-96828700 on Build GRCh38
rs747958442 Chr.2: 96828551-96828551 on Build GRCh38
rs767022280 Chr.2: 96828627-96828627 on Build GRCh38
rs374840487 Chr.2: 96828581-96828581 on Build GRCh38
rs149028425 Chr.2: 96828544-96828544 on Build GRCh38
rs756870572 Chr.2: 96828551-96828552 on Build GRCh38

View Full Product Details