Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 103724041-103724280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLRA4
Entrez Gene ID
441509
Gene Name
glycine receptor alpha 4
Gene Aliases
-
UniGene
Hs.533289
Gene Chromosome Location
Chr.X: 103707344-103728624 on Build GRCh38
Transcripts
NM_001024452.2 NM_001172285.1

SNP Detail

SNP ID Chromosome Location
rs371275422 Chr.X: 103724070-103724070 on Build GRCh38
rs564177682 Chr.X: 103724154-103724154 on Build GRCh38
rs773930441 Chr.X: 103724253-103724253 on Build GRCh38
rs185788563 Chr.X: 103724132-103724132 on Build GRCh38
rs190361501 Chr.X: 103724140-103724140 on Build GRCh38
rs750263049 Chr.X: 103724101-103724101 on Build GRCh38
rs200296619 Chr.X: 103724093-103724093 on Build GRCh38
rs763586970 Chr.X: 103724082-103724082 on Build GRCh38
rs775095283 Chr.X: 103724066-103724066 on Build GRCh38
rs752736748 Chr.X: 103724214-103724214 on Build GRCh38
rs995959548 Chr.X: 103724068-103724068 on Build GRCh38
rs370561650 Chr.X: 103724139-103724139 on Build GRCh38
rs761188279 Chr.X: 103724255-103724255 on Build GRCh38
rs775183174 Chr.X: 103724215-103724215 on Build GRCh38
rs747417815 Chr.X: 103724211-103724211 on Build GRCh38
rs16984014 Chr.X: 103724256-103724256 on Build GRCh38
rs765418444 Chr.X: 103724131-103724131 on Build GRCh38
rs201961966 Chr.X: 103724235-103724235 on Build GRCh38
rs750963784 Chr.X: 103724086-103724086 on Build GRCh38
rs1014977440 Chr.X: 103724110-103724110 on Build GRCh38
rs368100271 Chr.X: 103724209-103724209 on Build GRCh38
rs373227426 Chr.X: 103724071-103724071 on Build GRCh38
rs182137906 Chr.X: 103724259-103724259 on Build GRCh38
rs780471240 Chr.X: 103724115-103724115 on Build GRCh38
rs767303961 Chr.X: 103724099-103724099 on Build GRCh38
rs763674831 Chr.X: 103724245-103724245 on Build GRCh38
rs61749967 Chr.X: 103724114-103724114 on Build GRCh38
rs61732248 Chr.X: 103724167-103724167 on Build GRCh38
rs267606296 Chr.X: 103724145-103724145 on Build GRCh38

View Full Product Details