Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 143292296-143292523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP2
Entrez Gene ID
835
Gene Name
caspase 2
Gene Aliases
CASP-2, ICH1, NEDD-2, NEDD2, PPP1R57
UniGene
Hs.368982
Gene Chromosome Location
Chr.7: 143288215-143307696 on Build GRCh38
Transcripts
NM_001224.4 NM_032982.3 NM_032983.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3942094 c.318G>A p.L106L Chr.7: 143292392-143292392 on Build GRCh38
3832154 c.361A>C p.T121P Chr.7: 143292435-143292435 on Build GRCh38
4411551 c.364C>A p.L122I Chr.7: 143292438-143292438 on Build GRCh38
4849504 c.249G>C p.Q83H Chr.7: 143292323-143292323 on Build GRCh38
6620567 c.335G>A p.G112D Chr.7: 143292409-143292409 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs893357769 Chr.7: 143292475-143292475 on Build GRCh38
rs764648518 Chr.7: 143292351-143292351 on Build GRCh38
rs774984141 Chr.7: 143292468-143292468 on Build GRCh38
rs745514145 Chr.7: 143292458-143292458 on Build GRCh38
rs559973451 Chr.7: 143292484-143292484 on Build GRCh38
rs746876443 Chr.7: 143292440-143292440 on Build GRCh38
rs760770832 Chr.7: 143292394-143292395 on Build GRCh38
rs752010369 Chr.7: 143292377-143292377 on Build GRCh38
rs781208424 Chr.7: 143292323-143292323 on Build GRCh38
rs756289782 Chr.7: 143292414-143292414 on Build GRCh38
rs769448055 Chr.7: 143292460-143292460 on Build GRCh38
rs570907619 Chr.7: 143292429-143292429 on Build GRCh38
rs137989079 Chr.7: 143292467-143292467 on Build GRCh38
rs763400986 Chr.7: 143292335-143292335 on Build GRCh38
rs538096948 Chr.7: 143292464-143292464 on Build GRCh38
rs762351638 Chr.7: 143292471-143292471 on Build GRCh38
rs113837507 Chr.7: 143292437-143292437 on Build GRCh38
rs766396793 Chr.7: 143292419-143292419 on Build GRCh38
rs753903184 Chr.7: 143292423-143292423 on Build GRCh38
rs767906534 Chr.7: 143292403-143292403 on Build GRCh38
rs757036285 Chr.7: 143292455-143292455 on Build GRCh38
rs780908429 Chr.7: 143292456-143292456 on Build GRCh38
rs750685719 Chr.7: 143292404-143292404 on Build GRCh38
rs748009560 Chr.7: 143292466-143292466 on Build GRCh38
rs762263774 Chr.7: 143292388-143292388 on Build GRCh38
rs4647291 Chr.7: 143292495-143292495 on Build GRCh38

View Full Product Details