Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 83010174-83010435 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C15orf40
Entrez Gene ID
123207
Gene Name
chromosome 15 open reading frame 40
Gene Aliases
-
UniGene
Hs.352541
Gene Chromosome Location
Chr.15: 82988297-83012298 on Build GRCh38
Transcripts
NM_001160113.1 NM_001160114.1 NM_001160115.1 NM_001160116.1 NM_144597.2 NR_027649.1 NR_027650.1 XM_006720385.2 XM_011521212.2 XM_011521213.2 XM_011521214.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2012731 c.107T>G p.V36G Chr.15: 83010287-83010287 on Build GRCh38
2012727 c.156A>C p.T52T Chr.15: 83010238-83010238 on Build GRCh38
349117 c.55G>C p.E19Q Chr.15: 83010339-83010339 on Build GRCh38
1246511 c.152T>C p.V51A Chr.15: 83010242-83010242 on Build GRCh38
107715 c.120C>T p.I40I Chr.15: 83010274-83010274 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1002090929 Chr.15: 83010245-83010245 on Build GRCh38
rs766001761 Chr.15: 83010350-83010350 on Build GRCh38
rs781406754 Chr.15: 83010328-83010328 on Build GRCh38
rs763633665 Chr.15: 83010306-83010306 on Build GRCh38
rs772888120 Chr.15: 83010243-83010243 on Build GRCh38
rs775373535 Chr.15: 83010379-83010379 on Build GRCh38
rs748311916 Chr.15: 83010362-83010362 on Build GRCh38
rs371313767 Chr.15: 83010284-83010284 on Build GRCh38
rs774400039 Chr.15: 83010271-83010271 on Build GRCh38
rs762926374 Chr.15: 83010381-83010381 on Build GRCh38
rs772265696 Chr.15: 83010369-83010369 on Build GRCh38
rs762141610 Chr.15: 83010272-83010272 on Build GRCh38
rs905095153 Chr.15: 83010239-83010239 on Build GRCh38
rs770401805 Chr.15: 83010194-83010195 on Build GRCh38
rs780714622 Chr.15: 83010251-83010251 on Build GRCh38
rs757917041 Chr.15: 83010195-83010195 on Build GRCh38
rs755695302 Chr.15: 83010305-83010305 on Build GRCh38
rs751148895 Chr.15: 83010392-83010392 on Build GRCh38
rs756564956 Chr.15: 83010337-83010337 on Build GRCh38
rs763500837 Chr.15: 83010391-83010391 on Build GRCh38
rs746347805 Chr.15: 83010246-83010246 on Build GRCh38
rs942296307 Chr.15: 83010269-83010269 on Build GRCh38
rs781658936 Chr.15: 83010244-83010244 on Build GRCh38
rs149037136 Chr.15: 83010274-83010274 on Build GRCh38
rs777356088 Chr.15: 83010203-83010203 on Build GRCh38
rs116657382 Chr.15: 83010360-83010360 on Build GRCh38
rs1026511227 Chr.15: 83010322-83010322 on Build GRCh38
rs759276260 Chr.15: 83010371-83010371 on Build GRCh38
rs964923721 Chr.15: 83010383-83010383 on Build GRCh38
rs139912640 Chr.15: 83010249-83010249 on Build GRCh38
rs780663204 Chr.15: 83010339-83010339 on Build GRCh38
rs116595247 Chr.15: 83010334-83010334 on Build GRCh38
rs768882688 Chr.15: 83010254-83010254 on Build GRCh38
rs1042071827 Chr.15: 83010363-83010363 on Build GRCh38
rs114161234 Chr.15: 83010353-83010353 on Build GRCh38
rs769607952 Chr.15: 83010374-83010374 on Build GRCh38
rs367557181 Chr.15: 83010279-83010279 on Build GRCh38
rs752653882 Chr.15: 83010297-83010297 on Build GRCh38
rs757529839 Chr.15: 83010236-83010236 on Build GRCh38
rs779103561 Chr.15: 83010355-83010355 on Build GRCh38
rs746704252 Chr.15: 83010208-83010208 on Build GRCh38
rs773316651 Chr.15: 83010370-83010370 on Build GRCh38
rs143424937 Chr.15: 83010293-83010293 on Build GRCh38
rs749845710 Chr.15: 83010352-83010352 on Build GRCh38
rs751151578 Chr.15: 83010314-83010314 on Build GRCh38
rs1013285354 Chr.15: 83010361-83010361 on Build GRCh38
rs756993991 Chr.15: 83010323-83010323 on Build GRCh38
rs762519924 Chr.15: 83010334-83010337 on Build GRCh38

View Full Product Details