Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 182842016-182842247 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP11B
Entrez Gene ID
23200
Gene Name
ATPase phospholipid transporting 11B (putative)
Gene Aliases
ATPIF, ATPIR
UniGene
Hs.478429
Gene Chromosome Location
Chr.3: 182793503-182923010 on Build GRCh38
Transcripts
NM_014616.2 XM_005247241.2 XM_005247243.2 XM_011512593.2 XM_011512594.2 XM_011512597.2 XM_017006008.1 XM_017006009.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1753030 c.696A>T p.E232D Chr.3: 182842114-182842114 on Build GRCh38
3408458 c.665G>T p.G222V Chr.3: 182842083-182842083 on Build GRCh38
3846824 c.694G>T p.E232* Chr.3: 182842112-182842112 on Build GRCh38
6511800 c.664G>A p.G222R Chr.3: 182842082-182842082 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs2971454 Chr.3: 182842184-182842184 on Build GRCh38
rs35093142 Chr.3: 182842055-182842055 on Build GRCh38
rs1050556899 Chr.3: 182842126-182842126 on Build GRCh38
rs512215 Chr.3: 182842112-182842112 on Build GRCh38
rs34078217 Chr.3: 182842138-182842138 on Build GRCh38
rs761846502 Chr.3: 182842088-182842088 on Build GRCh38
rs750195934 Chr.3: 182842096-182842096 on Build GRCh38
rs767561810 Chr.3: 182842070-182842070 on Build GRCh38
rs769062591 Chr.3: 182842052-182842052 on Build GRCh38
rs903927274 Chr.3: 182842159-182842159 on Build GRCh38
rs976721536 Chr.3: 182842158-182842158 on Build GRCh38
rs765171647 Chr.3: 182842086-182842086 on Build GRCh38
rs770571152 Chr.3: 182842134-182842135 on Build GRCh38
rs758195994 Chr.3: 182842161-182842161 on Build GRCh38
rs372223959 Chr.3: 182842071-182842071 on Build GRCh38
rs773891343 Chr.3: 182842167-182842168 on Build GRCh38
rs751237870 Chr.3: 182842172-182842172 on Build GRCh38
rs551395741 Chr.3: 182842188-182842188 on Build GRCh38
rs766199091 Chr.3: 182842171-182842171 on Build GRCh38

View Full Product Details