Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15914793-15915020 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4F11
Entrez Gene ID
57834
Gene Name
cytochrome P450 family 4 subfamily F member 11
Gene Aliases
CYPIVF11
UniGene
Hs.187393
Gene Chromosome Location
Chr.19: 15912370-15934866 on Build GRCh38
Transcripts
NM_001128932.1 NM_021187.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3284293 c.1198C>T p.R400* Chr.19: 15914813-15914813 on Build GRCh38
6150237 c.1129C>A p.Q377K Chr.19: 15914882-15914882 on Build GRCh38
5614613 c.1214A>G p.D405G Chr.19: 15914797-15914797 on Build GRCh38
992373 c.1207A>G p.T403A Chr.19: 15914804-15914804 on Build GRCh38
1711972 c.1199G>A p.R400Q Chr.19: 15914812-15914812 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550369934 Chr.19: 15914906-15914906 on Build GRCh38
rs372437243 Chr.19: 15914813-15914813 on Build GRCh38
rs773559903 Chr.19: 15914884-15914884 on Build GRCh38
rs763634517 Chr.19: 15914830-15914830 on Build GRCh38
rs748298676 Chr.19: 15914947-15914947 on Build GRCh38
rs763829157 Chr.19: 15914897-15914897 on Build GRCh38
rs543811544 Chr.19: 15914907-15914912 on Build GRCh38
rs779083973 Chr.19: 15914943-15914943 on Build GRCh38
rs375164936 Chr.19: 15914905-15914905 on Build GRCh38
rs564152748 Chr.19: 15914968-15914968 on Build GRCh38
rs777895272 Chr.19: 15914910-15914910 on Build GRCh38
rs752351878 Chr.19: 15914825-15914825 on Build GRCh38
rs200976601 Chr.19: 15914918-15914918 on Build GRCh38
rs141887211 Chr.19: 15914824-15914824 on Build GRCh38
rs755823193 Chr.19: 15914844-15914844 on Build GRCh38
rs145791711 Chr.19: 15914879-15914879 on Build GRCh38
rs760483929 Chr.19: 15914952-15914952 on Build GRCh38
rs570135539 Chr.19: 15914912-15914912 on Build GRCh38
rs751168274 Chr.19: 15914908-15914908 on Build GRCh38
rs565985827 Chr.19: 15914969-15914969 on Build GRCh38
rs755947846 Chr.19: 15914917-15914917 on Build GRCh38
rs200588981 Chr.19: 15914812-15914812 on Build GRCh38
rs192392601 Chr.19: 15914970-15914970 on Build GRCh38
rs146262579 Chr.19: 15914892-15914892 on Build GRCh38
rs753851013 Chr.19: 15914901-15914901 on Build GRCh38
rs755250510 Chr.19: 15914940-15914940 on Build GRCh38
rs374763718 Chr.19: 15914843-15914843 on Build GRCh38
rs749450646 Chr.19: 15914862-15914862 on Build GRCh38
rs761200530 Chr.19: 15914909-15914909 on Build GRCh38
rs779030910 Chr.19: 15914871-15914871 on Build GRCh38
rs45574935 Chr.19: 15914841-15914841 on Build GRCh38
rs768723718 Chr.19: 15914864-15914864 on Build GRCh38
rs138369700 Chr.19: 15914823-15914823 on Build GRCh38
rs761126212 Chr.19: 15914891-15914891 on Build GRCh38
rs1004713453 Chr.19: 15914920-15914920 on Build GRCh38
rs567758136 Chr.19: 15914872-15914872 on Build GRCh38
rs558822665 Chr.19: 15914959-15914959 on Build GRCh38
rs753621794 Chr.19: 15914923-15914923 on Build GRCh38
rs201684723 Chr.19: 15914842-15914842 on Build GRCh38
rs780274514 Chr.19: 15914849-15914849 on Build GRCh38
rs767524132 Chr.19: 15914911-15914911 on Build GRCh38
rs762570221 Chr.19: 15914828-15914828 on Build GRCh38
rs754913399 Chr.19: 15914907-15914907 on Build GRCh38

View Full Product Details