Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57628061-57628318 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALNT1
Entrez Gene ID
2583
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 1
Gene Aliases
GALGT, GALNACT, GalNAc-T, SPG26
UniGene
Hs.159481
Gene Chromosome Location
Chr.12: 57623409-57633239 on Build GRCh38
Transcripts
NM_001276468.1 NM_001478.4 XM_005268773.4 XM_011538147.2 XM_011538148.2 XM_017019140.1 XM_017019141.1 XM_017019142.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4622699 c.1068C>T p.D356D Chr.12: 57628197-57628197 on Build GRCh38
6609384 c.1094C>T p.T365M Chr.12: 57628171-57628171 on Build GRCh38
6520385 c.1003G>T p.G335C Chr.12: 57628262-57628262 on Build GRCh38
4664917 c.1130C>T p.T377M Chr.12: 57628135-57628135 on Build GRCh38
6203802 c.1092G>T p.R364R Chr.12: 57628173-57628173 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772896451 Chr.12: 57628231-57628231 on Build GRCh38
rs763991428 Chr.12: 57628264-57628264 on Build GRCh38
rs762519517 Chr.12: 57628236-57628236 on Build GRCh38
rs759167970 Chr.12: 57628254-57628254 on Build GRCh38
rs747586827 Chr.12: 57628211-57628211 on Build GRCh38
rs913007874 Chr.12: 57628295-57628295 on Build GRCh38
rs1024057832 Chr.12: 57628246-57628246 on Build GRCh38
rs577024960 Chr.12: 57628098-57628098 on Build GRCh38
rs752528723 Chr.12: 57628260-57628260 on Build GRCh38
rs749350620 Chr.12: 57628170-57628170 on Build GRCh38
rs145474220 Chr.12: 57628171-57628171 on Build GRCh38
rs374808727 Chr.12: 57628110-57628110 on Build GRCh38
rs11616065 Chr.12: 57628241-57628241 on Build GRCh38
rs779042510 Chr.12: 57628082-57628082 on Build GRCh38
rs757352421 Chr.12: 57628273-57628273 on Build GRCh38
rs1018621107 Chr.12: 57628157-57628157 on Build GRCh38
rs745654266 Chr.12: 57628136-57628137 on Build GRCh38
rs144643461 Chr.12: 57628217-57628217 on Build GRCh38
rs775885487 Chr.12: 57628080-57628080 on Build GRCh38
rs901370463 Chr.12: 57628212-57628212 on Build GRCh38
rs767291126 Chr.12: 57628143-57628143 on Build GRCh38
rs746083828 Chr.12: 57628289-57628289 on Build GRCh38
rs202231637 Chr.12: 57628180-57628180 on Build GRCh38
rs756096629 Chr.12: 57628262-57628262 on Build GRCh38
rs925607866 Chr.12: 57628268-57628268 on Build GRCh38
rs753737342 Chr.12: 57628266-57628266 on Build GRCh38
rs779037403 Chr.12: 57628179-57628179 on Build GRCh38
rs752587364 Chr.12: 57628144-57628144 on Build GRCh38
rs61649746 Chr.12: 57628146-57628146 on Build GRCh38
rs541794502 Chr.12: 57628188-57628188 on Build GRCh38
rs12322482 Chr.12: 57628291-57628291 on Build GRCh38
rs961180352 Chr.12: 57628159-57628159 on Build GRCh38
rs144926023 Chr.12: 57628257-57628257 on Build GRCh38
rs775858514 Chr.12: 57628198-57628198 on Build GRCh38
rs367919141 Chr.12: 57628111-57628111 on Build GRCh38
rs777891652 Chr.12: 57628154-57628154 on Build GRCh38
rs754757482 Chr.12: 57628141-57628141 on Build GRCh38
rs771698791 Chr.12: 57628146-57628151 on Build GRCh38
rs751408839 Chr.12: 57628131-57628131 on Build GRCh38
rs982615765 Chr.12: 57628267-57628267 on Build GRCh38
rs1025757313 Chr.12: 57628225-57628225 on Build GRCh38
rs956573343 Chr.12: 57628112-57628112 on Build GRCh38
rs764844859 Chr.12: 57628107-57628107 on Build GRCh38
rs151177784 Chr.12: 57628130-57628130 on Build GRCh38
rs141521727 Chr.12: 57628279-57628279 on Build GRCh38

View Full Product Details