Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41619153-41619392 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEACAM4
Entrez Gene ID
1089
Gene Name
carcinoembryonic antigen related cell adhesion molecule 4
Gene Aliases
CGM7, CGM7_HUMAN, NCA
UniGene
Hs.12
Gene Chromosome Location
Chr.19: 41616986-41627087 on Build GRCh38
Transcripts
NM_001817.3 XM_006722977.3 XM_011526343.1 XM_011526345.2 XM_011526348.2 XM_011526349.2 XM_011526350.2 XM_011526352.1 XM_011526354.1 XM_011526357.2 XM_017026199.1 XM_017026200.1 XM_017026201.1 XM_017026202.1 XM_017026203.1 XM_017026204.1 XM_017026205.1 XM_017026206.1 XM_017026207.1 XM_017026208.1 XM_017026209.1 XM_017026210.1 XM_017026211.1 XM_017026212.1 XM_017026213.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7199689 c.688G>A p.A230T Chr.19: 41619377-41619377 on Build GRCh38
6150967 c.703_704CA>AT p.Q235M Chr.19: 41619361-41619362 on Build GRCh38
3748684 c.718G>T p.A240S Chr.19: 41619347-41619347 on Build GRCh38
4653144 c.723T>C p.D241D Chr.19: 41619342-41619342 on Build GRCh38
3892660 c.708C>T p.I236I Chr.19: 41619357-41619357 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782463139 Chr.19: 41619354-41619354 on Build GRCh38
rs951425417 Chr.19: 41619197-41619197 on Build GRCh38
rs139833578 Chr.19: 41619340-41619340 on Build GRCh38
rs948111665 Chr.19: 41619284-41619284 on Build GRCh38
rs782153195 Chr.19: 41619338-41619338 on Build GRCh38
rs782634640 Chr.19: 41619290-41619290 on Build GRCh38
rs782727802 Chr.19: 41619356-41619356 on Build GRCh38
rs891686134 Chr.19: 41619316-41619316 on Build GRCh38
rs782007228 Chr.19: 41619337-41619337 on Build GRCh38
rs199844441 Chr.19: 41619294-41619294 on Build GRCh38
rs371748343 Chr.19: 41619295-41619295 on Build GRCh38
rs782106093 Chr.19: 41619329-41619329 on Build GRCh38
rs192614863 Chr.19: 41619363-41619363 on Build GRCh38
rs1041994 Chr.19: 41619287-41619287 on Build GRCh38
rs974330244 Chr.19: 41619181-41619181 on Build GRCh38
rs8113662 Chr.19: 41619266-41619266 on Build GRCh38
rs148938635 Chr.19: 41619250-41619250 on Build GRCh38
rs1009862715 Chr.19: 41619331-41619331 on Build GRCh38
rs782332804 Chr.19: 41619311-41619311 on Build GRCh38
rs558445231 Chr.19: 41619176-41619176 on Build GRCh38
rs538937334 Chr.19: 41619267-41619267 on Build GRCh38
rs782765742 Chr.19: 41619232-41619232 on Build GRCh38
rs782768503 Chr.19: 41619336-41619336 on Build GRCh38
rs200632223 Chr.19: 41619293-41619293 on Build GRCh38
rs547573251 Chr.19: 41619274-41619274 on Build GRCh38
rs781791378 Chr.19: 41619341-41619341 on Build GRCh38
rs374967615 Chr.19: 41619296-41619296 on Build GRCh38
rs781958504 Chr.19: 41619317-41619317 on Build GRCh38
rs1041993 Chr.19: 41619289-41619289 on Build GRCh38
rs750572011 Chr.19: 41619182-41619182 on Build GRCh38

View Full Product Details