Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 154428021-154428294 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNKSR3
Entrez Gene ID
154043
Gene Name
CNKSR family member 3
Gene Aliases
MAGI1
UniGene
Hs.16064
Gene Chromosome Location
Chr.6: 154405299-154510619 on Build GRCh38
Transcripts
NM_173515.2 XM_011535485.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5184225 c.729+8T>C p.? Chr.6: 154428120-154428120 on Build GRCh38
160068 c.676T>C p.Y226H Chr.6: 154428181-154428181 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs540937233 Chr.6: 154428226-154428226 on Build GRCh38
rs376718197 Chr.6: 154428179-154428179 on Build GRCh38
rs535292357 Chr.6: 154428059-154428059 on Build GRCh38
rs1016415453 Chr.6: 154428072-154428072 on Build GRCh38
rs557464834 Chr.6: 154428198-154428198 on Build GRCh38
rs114854690 Chr.6: 154428245-154428245 on Build GRCh38
rs746213964 Chr.6: 154428227-154428227 on Build GRCh38
rs1022450673 Chr.6: 154428262-154428262 on Build GRCh38
rs758490671 Chr.6: 154428182-154428182 on Build GRCh38
rs761559497 Chr.6: 154428237-154428238 on Build GRCh38
rs369005792 Chr.6: 154428152-154428152 on Build GRCh38
rs746274275 Chr.6: 154428081-154428081 on Build GRCh38
rs933863988 Chr.6: 154428195-154428195 on Build GRCh38
rs370023781 Chr.6: 154428197-154428197 on Build GRCh38
rs778003393 Chr.6: 154428184-154428184 on Build GRCh38
rs372698348 Chr.6: 154428164-154428164 on Build GRCh38
rs538770220 Chr.6: 154428106-154428106 on Build GRCh38
rs1051013360 Chr.6: 154428214-154428214 on Build GRCh38
rs892282061 Chr.6: 154428243-154428243 on Build GRCh38
rs909325259 Chr.6: 154428046-154428046 on Build GRCh38
rs751755770 Chr.6: 154428186-154428186 on Build GRCh38
rs771594128 Chr.6: 154428121-154428121 on Build GRCh38
rs770088341 Chr.6: 154428237-154428237 on Build GRCh38
rs553403683 Chr.6: 154428062-154428062 on Build GRCh38
rs963089570 Chr.6: 154428087-154428087 on Build GRCh38
rs1007700718 Chr.6: 154428050-154428050 on Build GRCh38
rs567455356 Chr.6: 154428045-154428045 on Build GRCh38
rs773917618 Chr.6: 154428092-154428092 on Build GRCh38
rs9384217 Chr.6: 154428091-154428091 on Build GRCh38
rs766373683 Chr.6: 154428140-154428140 on Build GRCh38
rs753657137 Chr.6: 154428151-154428151 on Build GRCh38
rs761346974 Chr.6: 154428119-154428119 on Build GRCh38
rs760614924 Chr.6: 154428134-154428134 on Build GRCh38
rs959297567 Chr.6: 154428201-154428201 on Build GRCh38
rs773080032 Chr.6: 154428133-154428133 on Build GRCh38
rs1039076259 Chr.6: 154428123-154428123 on Build GRCh38

View Full Product Details