Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 22347627-22347870 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDR2
Entrez Gene ID
1039
Gene Name
cerebellar degeneration related protein 2
Gene Aliases
CDR62, Yo
UniGene
Hs.513430
Gene Chromosome Location
Chr.16: 22345936-22374617 on Build GRCh38
Transcripts
NM_001802.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3969454 c.577G>C p.E193Q Chr.16: 22347753-22347753 on Build GRCh38
3278769 c.638G>T p.R213L Chr.16: 22347692-22347692 on Build GRCh38
4552359 c.556G>A p.G186S Chr.16: 22347774-22347774 on Build GRCh38
3278770 c.601_602insA p.T201fs*32 Chr.16: 22347728-22347729 on Build GRCh38
3746400 c.667T>A p.Y223N Chr.16: 22347663-22347663 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200433383 Chr.16: 22347814-22347814 on Build GRCh38
rs372651445 Chr.16: 22347692-22347692 on Build GRCh38
rs774855594 Chr.16: 22347672-22347672 on Build GRCh38
rs974608275 Chr.16: 22347759-22347759 on Build GRCh38
rs557342634 Chr.16: 22347829-22347830 on Build GRCh38
rs113655596 Chr.16: 22347817-22347817 on Build GRCh38
rs373272674 Chr.16: 22347745-22347745 on Build GRCh38
rs768276956 Chr.16: 22347811-22347811 on Build GRCh38
rs371413069 Chr.16: 22347836-22347837 on Build GRCh38
rs769723609 Chr.16: 22347654-22347654 on Build GRCh38
rs751405114 Chr.16: 22347731-22347731 on Build GRCh38
rs779456514 Chr.16: 22347781-22347781 on Build GRCh38
rs377261960 Chr.16: 22347798-22347798 on Build GRCh38
rs972655941 Chr.16: 22347763-22347763 on Build GRCh38
rs758372907 Chr.16: 22347725-22347725 on Build GRCh38
rs955339090 Chr.16: 22347679-22347679 on Build GRCh38
rs777650420 Chr.16: 22347742-22347742 on Build GRCh38
rs749416692 Chr.16: 22347664-22347664 on Build GRCh38
rs765074249 Chr.16: 22347718-22347718 on Build GRCh38
rs369199367 Chr.16: 22347712-22347712 on Build GRCh38
rs756234545 Chr.16: 22347739-22347739 on Build GRCh38
rs372632705 Chr.16: 22347693-22347693 on Build GRCh38
rs774731079 Chr.16: 22347789-22347791 on Build GRCh38
rs766336966 Chr.16: 22347727-22347727 on Build GRCh38
rs146229941 Chr.16: 22347816-22347816 on Build GRCh38
rs746262316 Chr.16: 22347787-22347787 on Build GRCh38
rs760182484 Chr.16: 22347684-22347684 on Build GRCh38
rs772811607 Chr.16: 22347830-22347830 on Build GRCh38
rs776348664 Chr.16: 22347709-22347709 on Build GRCh38
rs748229057 Chr.16: 22347652-22347652 on Build GRCh38
rs750198394 Chr.16: 22347721-22347721 on Build GRCh38
rs372109741 Chr.16: 22347655-22347655 on Build GRCh38
rs772671638 Chr.16: 22347685-22347685 on Build GRCh38
rs776075174 Chr.16: 22347796-22347796 on Build GRCh38
rs776098459 Chr.16: 22347689-22347689 on Build GRCh38
rs747406634 Chr.16: 22347799-22347799 on Build GRCh38
rs771429999 Chr.16: 22347669-22347669 on Build GRCh38
rs767087379 Chr.16: 22347830-22347830 on Build GRCh38

View Full Product Details