Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6817279-6817548 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD4
Entrez Gene ID
920
Gene Name
CD4 molecule
Gene Aliases
CD4mut
UniGene
Hs.631659
Gene Chromosome Location
Chr.12: 6789472-6820810 on Build GRCh38
Transcripts
NM_000616.4 NM_001195014.2 NM_001195015.2 NM_001195016.2 NM_001195017.2 XM_017020228.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5882317 c.1149C>A p.N383K Chr.12: 6817323-6817323 on Build GRCh38
4921125 c.1125G>C p.S375S Chr.12: 6817299-6817299 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs782312850 Chr.12: 6817375-6817375 on Build GRCh38
rs982654538 Chr.12: 6817473-6817473 on Build GRCh38
rs782637634 Chr.12: 6817439-6817439 on Build GRCh38
rs200352608 Chr.12: 6817370-6817370 on Build GRCh38
rs782633339 Chr.12: 6817323-6817323 on Build GRCh38
rs764154464 Chr.12: 6817410-6817410 on Build GRCh38
rs200588692 Chr.12: 6817373-6817373 on Build GRCh38
rs782556070 Chr.12: 6817299-6817299 on Build GRCh38
rs782153257 Chr.12: 6817432-6817432 on Build GRCh38
rs201280358 Chr.12: 6817343-6817343 on Build GRCh38
rs782518852 Chr.12: 6817438-6817438 on Build GRCh38
rs200603669 Chr.12: 6817362-6817362 on Build GRCh38
rs751399122 Chr.12: 6817457-6817457 on Build GRCh38
rs757366202 Chr.12: 6817504-6817504 on Build GRCh38
rs782391302 Chr.12: 6817356-6817356 on Build GRCh38
rs113113481 Chr.12: 6817394-6817394 on Build GRCh38
rs781936005 Chr.12: 6817361-6817361 on Build GRCh38
rs782519691 Chr.12: 6817453-6817453 on Build GRCh38
rs578110262 Chr.12: 6817400-6817400 on Build GRCh38
rs199640148 Chr.12: 6817348-6817348 on Build GRCh38
rs11575103 Chr.12: 6817490-6817490 on Build GRCh38
rs782477319 Chr.12: 6817484-6817484 on Build GRCh38
rs781886210 Chr.12: 6817382-6817382 on Build GRCh38
rs200632431 Chr.12: 6817309-6817309 on Build GRCh38
rs3213426 Chr.12: 6817505-6817505 on Build GRCh38
rs199767708 Chr.12: 6817371-6817371 on Build GRCh38
rs991267326 Chr.12: 6817411-6817411 on Build GRCh38
rs781805059 Chr.12: 6817367-6817367 on Build GRCh38

View Full Product Details