Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 120394429-120394702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK5RAP2
Entrez Gene ID
55755
Gene Name
CDK5 regulatory subunit associated protein 2
Gene Aliases
C48, Cep215, MCPH3
UniGene
Hs.269560
Gene Chromosome Location
Chr.9: 120388869-120580170 on Build GRCh38
Transcripts
NM_001011649.2 NM_001272039.1 NM_018249.5 NR_073554.1 NR_073555.1 NR_073556.1 NR_073557.1 NR_073558.1 XM_006717182.1 XM_006717185.1 XM_011518860.1 XM_017014921.1 XM_017014922.1 XM_017014923.1 XM_017014924.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2151811 c.5528T>G p.M1843R Chr.9: 120394562-120394562 on Build GRCh38
3432814 c.5511G>T p.K1837N Chr.9: 120394579-120394579 on Build GRCh38
4919787 c.5486A>T p.H1829L Chr.9: 120394604-120394604 on Build GRCh38
1104629 c.5556G>T p.Q1852H Chr.9: 120394534-120394534 on Build GRCh38
5802439 c.5452-17_5454del20 p.? Chr.9: 120394636-120394655 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764357809 Chr.9: 120394466-120394466 on Build GRCh38
rs928385059 Chr.9: 120394457-120394457 on Build GRCh38
rs961893157 Chr.9: 120394507-120394507 on Build GRCh38
rs114128928 Chr.9: 120394636-120394636 on Build GRCh38
rs758443623 Chr.9: 120394496-120394496 on Build GRCh38
rs776715198 Chr.9: 120394584-120394584 on Build GRCh38
rs769433619 Chr.9: 120394538-120394538 on Build GRCh38
rs10984902 Chr.9: 120394464-120394464 on Build GRCh38
rs374625129 Chr.9: 120394573-120394573 on Build GRCh38
rs370525481 Chr.9: 120394489-120394489 on Build GRCh38
rs764085167 Chr.9: 120394580-120394580 on Build GRCh38
rs374958109 Chr.9: 120394668-120394668 on Build GRCh38
rs755755790 Chr.9: 120394646-120394646 on Build GRCh38
rs373083579 Chr.9: 120394490-120394490 on Build GRCh38
rs780111593 Chr.9: 120394499-120394499 on Build GRCh38
rs370378596 Chr.9: 120394644-120394644 on Build GRCh38
rs746197980 Chr.9: 120394558-120394558 on Build GRCh38
rs7859997 Chr.9: 120394452-120394452 on Build GRCh38
rs757429869 Chr.9: 120394479-120394479 on Build GRCh38
rs774603556 Chr.9: 120394539-120394539 on Build GRCh38
rs746092997 Chr.9: 120394454-120394454 on Build GRCh38
rs746570860 Chr.9: 120394503-120394503 on Build GRCh38
rs760807024 Chr.9: 120394574-120394574 on Build GRCh38
rs531877703 Chr.9: 120394634-120394634 on Build GRCh38
rs759058853 Chr.9: 120394551-120394551 on Build GRCh38
rs200816141 Chr.9: 120394463-120394463 on Build GRCh38
rs772613222 Chr.9: 120394567-120394567 on Build GRCh38
rs775760128 Chr.9: 120394665-120394665 on Build GRCh38
rs766482413 Chr.9: 120394623-120394623 on Build GRCh38
rs750226842 Chr.9: 120394605-120394605 on Build GRCh38
rs367948561 Chr.9: 120394671-120394671 on Build GRCh38
rs567642243 Chr.9: 120394610-120394610 on Build GRCh38
rs4410972 Chr.9: 120394483-120394483 on Build GRCh38
rs751538526 Chr.9: 120394630-120394630 on Build GRCh38
rs747981409 Chr.9: 120394503-120394504 on Build GRCh38
rs371368446 Chr.9: 120394530-120394530 on Build GRCh38
rs145708139 Chr.9: 120394585-120394585 on Build GRCh38
rs765457208 Chr.9: 120394604-120394604 on Build GRCh38
rs766934798 Chr.9: 120394667-120394667 on Build GRCh38
rs772417533 Chr.9: 120394656-120394656 on Build GRCh38
rs754002855 Chr.9: 120394476-120394476 on Build GRCh38
rs189396476 Chr.9: 120394591-120394591 on Build GRCh38
rs77100552 Chr.9: 120394512-120394512 on Build GRCh38
rs777456961 Chr.9: 120394649-120394649 on Build GRCh38

View Full Product Details