Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27223506-27223777 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAD
Entrez Gene ID
790
Gene Name
carbamoyl-phosphate synthetase 2, aspartate transcarbamylase, and dihydroorotase
Gene Aliases
CDG1Z
UniGene
Hs.377010
Gene Chromosome Location
Chr.2: 27217257-27243792 on Build GRCh38
Transcripts
NM_001306079.1 NM_004341.4 XM_005264555.3 XM_006712101.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5814456 c.994A>T p.S332C Chr.2: 27223747-27223747 on Build GRCh38
13846 c.965G>T p.G322V Chr.2: 27223718-27223718 on Build GRCh38
395240 c.986C>T p.P329L Chr.2: 27223739-27223739 on Build GRCh38
6091016 c.870G>A p.L290L Chr.2: 27223623-27223623 on Build GRCh38
5511698 c.942C>T p.N314N Chr.2: 27223695-27223695 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746015748 Chr.2: 27223689-27223689 on Build GRCh38
rs774492044 Chr.2: 27223681-27223681 on Build GRCh38
rs753478495 Chr.2: 27223612-27223612 on Build GRCh38
rs142361149 Chr.2: 27223696-27223696 on Build GRCh38
rs1056956031 Chr.2: 27223734-27223734 on Build GRCh38
rs150192801 Chr.2: 27223714-27223714 on Build GRCh38
rs920328473 Chr.2: 27223530-27223530 on Build GRCh38
rs771018227 Chr.2: 27223557-27223557 on Build GRCh38
rs774435361 Chr.2: 27223559-27223559 on Build GRCh38
rs754943602 Chr.2: 27223570-27223571 on Build GRCh38
rs773351400 Chr.2: 27223533-27223533 on Build GRCh38
rs142425046 Chr.2: 27223607-27223607 on Build GRCh38
rs768018252 Chr.2: 27223580-27223580 on Build GRCh38
rs753199599 Chr.2: 27223587-27223587 on Build GRCh38
rs764471966 Chr.2: 27223711-27223711 on Build GRCh38
rs776337242 Chr.2: 27223700-27223700 on Build GRCh38
rs756969236 Chr.2: 27223613-27223613 on Build GRCh38
rs761004094 Chr.2: 27223603-27223603 on Build GRCh38
rs781071282 Chr.2: 27223653-27223653 on Build GRCh38
rs749955801 Chr.2: 27223615-27223615 on Build GRCh38
rs763202712 Chr.2: 27223544-27223544 on Build GRCh38
rs147119054 Chr.2: 27223626-27223626 on Build GRCh38
rs778383391 Chr.2: 27223614-27223614 on Build GRCh38
rs532422388 Chr.2: 27223747-27223747 on Build GRCh38
rs890088134 Chr.2: 27223600-27223600 on Build GRCh38
rs976184515 Chr.2: 27223543-27223543 on Build GRCh38
rs768375609 Chr.2: 27223652-27223652 on Build GRCh38
rs761218212 Chr.2: 27223705-27223705 on Build GRCh38
rs1050043944 Chr.2: 27223576-27223576 on Build GRCh38
rs201395949 Chr.2: 27223658-27223658 on Build GRCh38
rs746916632 Chr.2: 27223640-27223640 on Build GRCh38
rs377491157 Chr.2: 27223755-27223755 on Build GRCh38
rs868779862 Chr.2: 27223707-27223707 on Build GRCh38
rs942059036 Chr.2: 27223654-27223655 on Build GRCh38
rs759552111 Chr.2: 27223573-27223573 on Build GRCh38
rs780194881 Chr.2: 27223627-27223627 on Build GRCh38
rs867114012 Chr.2: 27223667-27223667 on Build GRCh38
rs543954596 Chr.2: 27223712-27223712 on Build GRCh38
rs938598214 Chr.2: 27223722-27223722 on Build GRCh38
rs929365599 Chr.2: 27223566-27223566 on Build GRCh38
rs373528595 Chr.2: 27223754-27223754 on Build GRCh38
rs891662391 Chr.2: 27223735-27223735 on Build GRCh38
rs368678097 Chr.2: 27223720-27223720 on Build GRCh38
rs369713312 Chr.2: 27223695-27223695 on Build GRCh38
rs771196504 Chr.2: 27223662-27223662 on Build GRCh38
rs764497829 Chr.2: 27223606-27223606 on Build GRCh38

View Full Product Details