Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 34849890-34850139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGT7
Entrez Gene ID
2686
Gene Name
gamma-glutamyltransferase 7
Gene Aliases
D20S101, GGT4, GGTL3, GGTL5, dJ18C9.2
UniGene
Hs.433738
Gene Chromosome Location
Chr.20: 34844720-34872858 on Build GRCh38
Transcripts
NM_178026.2 XM_011528782.2 XM_011528783.2 XM_017027809.1 XM_017027810.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4766839 c.1753C>T p.R585W Chr.20: 34850033-34850033 on Build GRCh38
1208151 c.1778G>A p.R593H Chr.20: 34850008-34850008 on Build GRCh38
4097820 c.1791C>G p.H597Q Chr.20: 34849995-34849995 on Build GRCh38
185775 c.1794G>A p.P598P Chr.20: 34849992-34849992 on Build GRCh38
4823512 c.1815G>T p.L605L Chr.20: 34849971-34849971 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750240150 Chr.20: 34850068-34850068 on Build GRCh38
rs759672583 Chr.20: 34850091-34850091 on Build GRCh38
rs781021139 Chr.20: 34849938-34849938 on Build GRCh38
rs758779721 Chr.20: 34849939-34849939 on Build GRCh38
rs375464567 Chr.20: 34849992-34849992 on Build GRCh38
rs200416866 Chr.20: 34850032-34850032 on Build GRCh38
rs759791217 Chr.20: 34850015-34850015 on Build GRCh38
rs1053536303 Chr.20: 34849984-34849984 on Build GRCh38
rs778279737 Chr.20: 34850004-34850004 on Build GRCh38
rs755889670 Chr.20: 34850074-34850074 on Build GRCh38
rs1009272224 Chr.20: 34850002-34850002 on Build GRCh38
rs371023655 Chr.20: 34850107-34850107 on Build GRCh38
rs1001938186 Chr.20: 34849948-34849948 on Build GRCh38
rs374099114 Chr.20: 34850094-34850094 on Build GRCh38
rs776955618 Chr.20: 34850011-34850011 on Build GRCh38
rs368879321 Chr.20: 34849993-34849993 on Build GRCh38
rs763031423 Chr.20: 34850026-34850026 on Build GRCh38
rs553428261 Chr.20: 34849941-34849941 on Build GRCh38
rs755799106 Chr.20: 34849942-34849942 on Build GRCh38
rs967645494 Chr.20: 34850006-34850006 on Build GRCh38
rs753296864 Chr.20: 34850089-34850089 on Build GRCh38
rs140458732 Chr.20: 34850007-34850007 on Build GRCh38
rs771238698 Chr.20: 34850008-34850008 on Build GRCh38
rs767462340 Chr.20: 34850045-34850045 on Build GRCh38
rs751325145 Chr.20: 34849933-34849933 on Build GRCh38
rs764079719 Chr.20: 34849932-34849932 on Build GRCh38
rs751472295 Chr.20: 34850033-34850033 on Build GRCh38
rs377626254 Chr.20: 34850103-34850103 on Build GRCh38
rs772474622 Chr.20: 34850003-34850003 on Build GRCh38
rs747567260 Chr.20: 34850102-34850102 on Build GRCh38
rs765346401 Chr.20: 34849911-34849911 on Build GRCh38
rs760756280 Chr.20: 34849934-34849934 on Build GRCh38
rs775486884 Chr.20: 34850025-34850025 on Build GRCh38
rs769995440 Chr.20: 34850017-34850017 on Build GRCh38
rs758331025 Chr.20: 34849931-34849931 on Build GRCh38
rs752788147 Chr.20: 34849918-34849918 on Build GRCh38
rs769921656 Chr.20: 34850108-34850108 on Build GRCh38
rs1015864072 Chr.20: 34849978-34849978 on Build GRCh38
rs370887247 Chr.20: 34850078-34850078 on Build GRCh38

View Full Product Details