Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 85804934-85805165 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX4I1
Entrez Gene ID
1327
Gene Name
cytochrome c oxidase subunit 4I1
Gene Aliases
COX IV-1, COX4, COX4-1, COXIV, COXIV-1
UniGene
Hs.433419
Gene Chromosome Location
Chr.16: 85799567-85807003 on Build GRCh38
Transcripts
NM_001318786.1 NM_001318788.1 NM_001318794.1 NM_001318797.1 NM_001318802.1 NM_001861.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
974529 c.90C>T p.S30S Chr.16: 85804953-85804953 on Build GRCh38
23302 c.117G>A p.M39I Chr.16: 85804980-85804980 on Build GRCh38
974531 c.121C>A p.R41R Chr.16: 85804984-85804984 on Build GRCh38
5385510 c.150C>T p.A50A Chr.16: 85805013-85805013 on Build GRCh38
5633061 c.136T>C p.L46L Chr.16: 85804999-85804999 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755854485 Chr.16: 85805107-85805107 on Build GRCh38
rs370929782 Chr.16: 85805017-85805017 on Build GRCh38
rs747411276 Chr.16: 85805074-85805074 on Build GRCh38
rs755549888 Chr.16: 85805022-85805022 on Build GRCh38
rs887346101 Chr.16: 85805078-85805078 on Build GRCh38
rs377678815 Chr.16: 85804965-85804965 on Build GRCh38
rs377718678 Chr.16: 85805132-85805132 on Build GRCh38
rs900925513 Chr.16: 85805112-85805112 on Build GRCh38
rs758936363 Chr.16: 85805121-85805121 on Build GRCh38
rs780684895 Chr.16: 85804978-85804978 on Build GRCh38
rs762932616 Chr.16: 85805004-85805004 on Build GRCh38
rs769159647 Chr.16: 85804984-85804984 on Build GRCh38
rs771139213 Chr.16: 85805075-85805075 on Build GRCh38
rs759508701 Chr.16: 85804996-85804996 on Build GRCh38
rs201058411 Chr.16: 85804964-85804964 on Build GRCh38
rs2599091 Chr.16: 85805047-85805047 on Build GRCh38
rs753408068 Chr.16: 85805120-85805120 on Build GRCh38
rs199978628 Chr.16: 85805100-85805100 on Build GRCh38
rs561331286 Chr.16: 85805086-85805086 on Build GRCh38
rs142400268 Chr.16: 85805103-85805103 on Build GRCh38
rs746164704 Chr.16: 85805082-85805082 on Build GRCh38
rs756915909 Chr.16: 85804971-85804971 on Build GRCh38
rs780067773 Chr.16: 85805145-85805145 on Build GRCh38
rs749391143 Chr.16: 85805146-85805146 on Build GRCh38
rs1056208673 Chr.16: 85804986-85804986 on Build GRCh38
rs368486192 Chr.16: 85805083-85805083 on Build GRCh38
rs774416869 Chr.16: 85805097-85805097 on Build GRCh38
rs771065408 Chr.16: 85804992-85804992 on Build GRCh38
rs751359351 Chr.16: 85805010-85805010 on Build GRCh38
rs778449832 Chr.16: 85805122-85805122 on Build GRCh38
rs765105753 Chr.16: 85804960-85804960 on Build GRCh38
rs764048670 Chr.16: 85805094-85805094 on Build GRCh38
rs1048702246 Chr.16: 85805073-85805073 on Build GRCh38
rs778220898 Chr.16: 85805065-85805065 on Build GRCh38
rs756860726 Chr.16: 85805014-85805014 on Build GRCh38
rs773110239 Chr.16: 85805041-85805041 on Build GRCh38
rs747556114 Chr.16: 85805125-85805125 on Build GRCh38
rs770277920 Chr.16: 85804994-85804995 on Build GRCh38
rs781661744 Chr.16: 85805130-85805130 on Build GRCh38
rs757556403 Chr.16: 85805129-85805129 on Build GRCh38
rs746016496 Chr.16: 85805131-85805131 on Build GRCh38
rs149461099 Chr.16: 85804985-85804985 on Build GRCh38
rs761492668 Chr.16: 85805101-85805101 on Build GRCh38
rs758991267 Chr.16: 85805048-85805048 on Build GRCh38
rs201313443 Chr.16: 85804988-85804988 on Build GRCh38
rs750140438 Chr.16: 85805019-85805019 on Build GRCh38
rs376218422 Chr.16: 85805087-85805087 on Build GRCh38
rs991810828 Chr.16: 85805009-85805009 on Build GRCh38
rs751199603 Chr.16: 85804968-85804968 on Build GRCh38
rs772561167 Chr.16: 85804987-85804987 on Build GRCh38
rs11557185 Chr.16: 85804975-85804975 on Build GRCh38
rs745400280 Chr.16: 85804980-85804980 on Build GRCh38
rs539978958 Chr.16: 85804963-85804963 on Build GRCh38
rs776735855 Chr.16: 85804995-85804995 on Build GRCh38
rs375329322 Chr.16: 85805114-85805114 on Build GRCh38
rs4885 Chr.16: 85805091-85805091 on Build GRCh38
rs932067795 Chr.16: 85804973-85804973 on Build GRCh38
rs775398027 Chr.16: 85805003-85805003 on Build GRCh38
rs773763510 Chr.16: 85805048-85805049 on Build GRCh38
rs770498990 Chr.16: 85804997-85804997 on Build GRCh38
rs913801825 Chr.16: 85804957-85804957 on Build GRCh38

View Full Product Details