Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 126464787-126465024 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf90
Entrez Gene ID
118611
Gene Name
chromosome 10 open reading frame 90
Gene Aliases
FATS, bA422P15.2
UniGene
Hs.587663
Gene Chromosome Location
Chr.10: 126424997-126671038 on Build GRCh38
Transcripts
NM_001004298.2 XM_005269491.1 XM_005269492.2 XM_011539216.1 XM_011539217.2 XM_011539220.2 XM_011539222.2 XM_011539223.2 XM_011539224.2 XM_011539226.1 XM_011539227.2 XM_011539230.2 XM_011539231.1 XM_011539232.1 XM_011539233.1 XM_017015634.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4873355 c.1695C>A p.P565P Chr.10: 126464826-126464826 on Build GRCh38
5915913 c.1598C>T p.S533F Chr.10: 126464923-126464923 on Build GRCh38
5899007 c.1640C>T p.P547L Chr.10: 126464881-126464881 on Build GRCh38
4416390 c.1601C>A p.A534D Chr.10: 126464920-126464920 on Build GRCh38
5727618 c.1535-1G>A p.? Chr.10: 126464987-126464987 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758287530 Chr.10: 126464906-126464906 on Build GRCh38
rs149443145 Chr.10: 126464916-126464916 on Build GRCh38
rs765912475 Chr.10: 126464886-126464886 on Build GRCh38
rs373175630 Chr.10: 126464921-126464921 on Build GRCh38
rs930696769 Chr.10: 126464835-126464835 on Build GRCh38
rs775472544 Chr.10: 126464818-126464818 on Build GRCh38
rs549988140 Chr.10: 126464935-126464935 on Build GRCh38
rs138712008 Chr.10: 126464863-126464863 on Build GRCh38
rs761130977 Chr.10: 126464882-126464882 on Build GRCh38
rs777794117 Chr.10: 126464813-126464813 on Build GRCh38
rs977962064 Chr.10: 126464939-126464939 on Build GRCh38
rs768922256 Chr.10: 126464851-126464851 on Build GRCh38
rs924674363 Chr.10: 126464945-126464945 on Build GRCh38
rs752201184 Chr.10: 126464942-126464944 on Build GRCh38
rs778709567 Chr.10: 126464900-126464900 on Build GRCh38
rs114882604 Chr.10: 126464816-126464816 on Build GRCh38
rs201855791 Chr.10: 126464902-126464902 on Build GRCh38
rs765173646 Chr.10: 126464863-126464873 on Build GRCh38
rs763231612 Chr.10: 126464847-126464847 on Build GRCh38
rs138339443 Chr.10: 126464815-126464815 on Build GRCh38
rs201947519 Chr.10: 126464911-126464911 on Build GRCh38
rs966925442 Chr.10: 126464936-126464936 on Build GRCh38
rs746933496 Chr.10: 126464912-126464912 on Build GRCh38
rs563866378 Chr.10: 126464899-126464899 on Build GRCh38
rs1004386819 Chr.10: 126464828-126464828 on Build GRCh38
rs74814928 Chr.10: 126464859-126464859 on Build GRCh38
rs199706107 Chr.10: 126464891-126464891 on Build GRCh38
rs774633302 Chr.10: 126464855-126464855 on Build GRCh38
rs369045364 Chr.10: 126464930-126464930 on Build GRCh38
rs1054456362 Chr.10: 126464979-126464979 on Build GRCh38
rs936121805 Chr.10: 126464946-126464946 on Build GRCh38

View Full Product Details