Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 186276328-186276569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F11
Entrez Gene ID
2160
Gene Name
coagulation factor XI
Gene Aliases
FXI
UniGene
Hs.1430
Gene Chromosome Location
Chr.4: 186265945-186290727 on Build GRCh38
Transcripts
NM_000128.3 XM_005262821.3 XM_005262822.3 XM_005262823.3 XM_006714137.2 XM_017007884.1 XM_017007885.1 XM_017007886.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7103509 c.751C>G p.Q251E Chr.4: 186276386-186276386 on Build GRCh38
4123834 c.716T>G p.F239C Chr.4: 186276351-186276351 on Build GRCh38
7103510 c.595C>G p.Q199E Chr.4: 186276386-186276386 on Build GRCh38
3602606 c.546C>T p.P182P Chr.4: 186276337-186276337 on Build GRCh38
1053943 c.745G>T p.E249* Chr.4: 186276380-186276380 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1041895911 Chr.4: 186276450-186276450 on Build GRCh38
rs573102359 Chr.4: 186276524-186276524 on Build GRCh38
rs281875260 Chr.4: 186276390-186276390 on Build GRCh38
rs750872465 Chr.4: 186276430-186276430 on Build GRCh38
rs989707969 Chr.4: 186276544-186276544 on Build GRCh38
rs762813060 Chr.4: 186276368-186276368 on Build GRCh38
rs894486730 Chr.4: 186276528-186276528 on Build GRCh38
rs987542299 Chr.4: 186276506-186276506 on Build GRCh38
rs533952931 Chr.4: 186276483-186276483 on Build GRCh38
rs281875279 Chr.4: 186276373-186276373 on Build GRCh38
rs747702749 Chr.4: 186276365-186276365 on Build GRCh38
rs1030979898 Chr.4: 186276507-186276507 on Build GRCh38
rs757275646 Chr.4: 186276391-186276391 on Build GRCh38
rs758898387 Chr.4: 186276434-186276434 on Build GRCh38
rs765242225 Chr.4: 186276422-186276422 on Build GRCh38
rs995963085 Chr.4: 186276369-186276369 on Build GRCh38
rs281875265 Chr.4: 186276358-186276358 on Build GRCh38
rs554127345 Chr.4: 186276505-186276505 on Build GRCh38
rs764339346 Chr.4: 186276384-186276384 on Build GRCh38
rs754087775 Chr.4: 186276386-186276386 on Build GRCh38
rs780457850 Chr.4: 186276441-186276441 on Build GRCh38
rs768484910 Chr.4: 186276356-186276356 on Build GRCh38
rs569813011 Chr.4: 186276541-186276541 on Build GRCh38
rs946255003 Chr.4: 186276399-186276399 on Build GRCh38
rs568171348 Chr.4: 186276374-186276374 on Build GRCh38
rs904641152 Chr.4: 186276479-186276479 on Build GRCh38
rs1014640590 Chr.4: 186276543-186276543 on Build GRCh38
rs1028819220 Chr.4: 186276452-186276452 on Build GRCh38
rs773153179 Chr.4: 186276367-186276367 on Build GRCh38
rs5969 Chr.4: 186276366-186276366 on Build GRCh38

View Full Product Details