Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 110376017-110376282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC7
Entrez Gene ID
51434
Gene Name
anaphase promoting complex subunit 7
Gene Aliases
APC7
UniGene
Hs.719935
Gene Chromosome Location
Chr.12: 110372900-110403730 on Build GRCh38
Transcripts
NM_001137664.1 NM_016238.2 XM_005253903.3 XM_011538454.2 XM_017019425.1 XM_017019426.1 XM_017019427.1 XM_017019428.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6599575 c.1503C>T p.N501N Chr.12: 110376173-110376173 on Build GRCh38
5840092 c.1481C>A p.A494E Chr.12: 110376093-110376093 on Build GRCh38
3810977 c.1497G>A p.L499L Chr.12: 110376179-110376179 on Build GRCh38
5889184 c.1460-7C>T p.? Chr.12: 110376223-110376223 on Build GRCh38
1586164 c.1510G>A p.A504T Chr.12: 110376166-110376166 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372651086 Chr.12: 110376118-110376118 on Build GRCh38
rs984158287 Chr.12: 110376048-110376048 on Build GRCh38
rs769894816 Chr.12: 110376138-110376138 on Build GRCh38
rs909946494 Chr.12: 110376054-110376054 on Build GRCh38
rs200361388 Chr.12: 110376137-110376137 on Build GRCh38
rs375415723 Chr.12: 110376221-110376221 on Build GRCh38
rs375690201 Chr.12: 110376187-110376187 on Build GRCh38
rs371110033 Chr.12: 110376211-110376211 on Build GRCh38
rs939569596 Chr.12: 110376172-110376172 on Build GRCh38
rs752662978 Chr.12: 110376104-110376104 on Build GRCh38
rs767659183 Chr.12: 110376091-110376091 on Build GRCh38
rs1037479987 Chr.12: 110376174-110376174 on Build GRCh38
rs766754223 Chr.12: 110376050-110376050 on Build GRCh38
rs759818197 Chr.12: 110376249-110376249 on Build GRCh38
rs757598375 Chr.12: 110376131-110376131 on Build GRCh38
rs757928328 Chr.12: 110376173-110376173 on Build GRCh38
rs777343991 Chr.12: 110376254-110376255 on Build GRCh38
rs969522109 Chr.12: 110376058-110376058 on Build GRCh38
rs759639186 Chr.12: 110376080-110376080 on Build GRCh38
rs761687318 Chr.12: 110376046-110376046 on Build GRCh38
rs768772375 Chr.12: 110376213-110376213 on Build GRCh38
rs981321716 Chr.12: 110376063-110376063 on Build GRCh38
rs879027791 Chr.12: 110376039-110376039 on Build GRCh38
rs769662800 Chr.12: 110376232-110376232 on Build GRCh38
rs772836382 Chr.12: 110376240-110376240 on Build GRCh38
rs774600740 Chr.12: 110376059-110376059 on Build GRCh38
rs772119984 Chr.12: 110376255-110376255 on Build GRCh38
rs764584620 Chr.12: 110376112-110376112 on Build GRCh38
rs779616373 Chr.12: 110376180-110376180 on Build GRCh38
rs951359810 Chr.12: 110376047-110376047 on Build GRCh38

View Full Product Details