Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 48759123-48759348 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGBL4
Entrez Gene ID
84871
Gene Name
ATP/GTP binding protein like 4
Gene Aliases
CCP6
UniGene
Hs.551844
Gene Chromosome Location
Chr.1: 48520157-50024011 on Build GRCh38
Transcripts
NM_001323573.1 NM_001323574.1 NM_001323575.1 NM_032785.3 NR_136623.1 XM_011542308.2 XM_017002595.1 XM_017002596.1 XM_017002597.1 NM_001302082.1 NM_024603.3 XM_011542141.2 XM_011542142.2 XM_017002331.1 XM_017002332.1 XM_017002333.1 XM_017002334.1

Gene Symbol
BEND5
Entrez Gene ID
79656
Gene Name
BEN domain containing 5
Gene Aliases
C1orf165
UniGene
Hs.475348
Gene Chromosome Location
Chr.1: 48727523-48778603 on Build GRCh38
Transcripts
NM_001323573.1 NM_001323574.1 NM_001323575.1 NM_032785.3 NR_136623.1 XM_011542308.2 XM_017002595.1 XM_017002596.1 XM_017002597.1 NM_001302082.1 NM_024603.3 XM_011542141.2 XM_011542142.2 XM_017002331.1 XM_017002332.1 XM_017002333.1 XM_017002334.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7293608 c.5A>T p.D2V Chr.1: 48759133-48759133 on Build GRCh38
6200258 c.386A>C p.E129A Chr.1: 48759259-48759259 on Build GRCh38
5629643 c.364C>A p.P122T Chr.1: 48759281-48759281 on Build GRCh38
6191816 c.402C>T p.S134S Chr.1: 48759243-48759243 on Build GRCh38
7293607 c.512A>T p.D171V Chr.1: 48759133-48759133 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367903944 Chr.1: 48759148-48759148 on Build GRCh38
rs777325753 Chr.1: 48759150-48759150 on Build GRCh38
rs773513345 Chr.1: 48759183-48759183 on Build GRCh38
rs904645045 Chr.1: 48759223-48759223 on Build GRCh38
rs1042186251 Chr.1: 48759219-48759219 on Build GRCh38
rs772557186 Chr.1: 48759311-48759311 on Build GRCh38
rs962471023 Chr.1: 48759253-48759253 on Build GRCh38
rs780671877 Chr.1: 48759246-48759246 on Build GRCh38
rs774501741 Chr.1: 48759170-48759170 on Build GRCh38
rs745604168 Chr.1: 48759154-48759154 on Build GRCh38
rs201623671 Chr.1: 48759206-48759206 on Build GRCh38
rs202108114 Chr.1: 48759190-48759190 on Build GRCh38
rs760919716 Chr.1: 48759184-48759184 on Build GRCh38
rs540778259 Chr.1: 48759158-48759158 on Build GRCh38
rs531139897 Chr.1: 48759298-48759298 on Build GRCh38
rs748438575 Chr.1: 48759265-48759265 on Build GRCh38
rs374150750 Chr.1: 48759164-48759164 on Build GRCh38
rs748136013 Chr.1: 48759271-48759271 on Build GRCh38
rs773286114 Chr.1: 48759299-48759299 on Build GRCh38
rs557411763 Chr.1: 48759252-48759252 on Build GRCh38
rs1023747124 Chr.1: 48759247-48759247 on Build GRCh38
rs749096188 Chr.1: 48759264-48759264 on Build GRCh38
rs56982079 Chr.1: 48759314-48759314 on Build GRCh38
rs776914684 Chr.1: 48759315-48759315 on Build GRCh38
rs999900027 Chr.1: 48759156-48759156 on Build GRCh38
rs767585722 Chr.1: 48759178-48759178 on Build GRCh38
rs761016220 Chr.1: 48759307-48759307 on Build GRCh38
rs759911695 Chr.1: 48759318-48759318 on Build GRCh38
rs779216654 Chr.1: 48759207-48759207 on Build GRCh38
rs780057963 Chr.1: 48759261-48759261 on Build GRCh38
rs56960839 Chr.1: 48759313-48759313 on Build GRCh38
rs756896304 Chr.1: 48759177-48759177 on Build GRCh38
rs750979533 Chr.1: 48759239-48759239 on Build GRCh38
rs768848202 Chr.1: 48759165-48759165 on Build GRCh38
rs148968303 Chr.1: 48759153-48759153 on Build GRCh38
rs765743070 Chr.1: 48759232-48759232 on Build GRCh38
rs746673782 Chr.1: 48759151-48759151 on Build GRCh38
rs755957106 Chr.1: 48759255-48759255 on Build GRCh38

View Full Product Details