Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 115942305-115942570 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRK
Entrez Gene ID
2444
Gene Name
fyn related Src family tyrosine kinase
Gene Aliases
GTK, PTK5, RAK
UniGene
Hs.89426
Gene Chromosome Location
Chr.6: 115941031-116100739 on Build GRCh38
Transcripts
NM_002031.2 XM_005266880.4 XM_005266881.2 XM_005266882.4 XM_011535653.2 XM_011535654.2 XM_011535655.2 XM_011535656.2 XM_017010645.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1072336 c.1451G>A p.R484H Chr.6: 115942481-115942481 on Build GRCh38
74702 c.1450C>T p.R484C Chr.6: 115942482-115942482 on Build GRCh38
739573 c.1451G>T p.R484L Chr.6: 115942481-115942481 on Build GRCh38
5803907 c.1379A>T p.Q460L Chr.6: 115942553-115942553 on Build GRCh38
5869136 c.1505A>G p.N502S Chr.6: 115942427-115942427 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148760208 Chr.6: 115942459-115942459 on Build GRCh38
rs563923955 Chr.6: 115942430-115942430 on Build GRCh38
rs774189980 Chr.6: 115942414-115942415 on Build GRCh38
rs761637626 Chr.6: 115942517-115942517 on Build GRCh38
rs142072444 Chr.6: 115942496-115942496 on Build GRCh38
rs141525046 Chr.6: 115942481-115942481 on Build GRCh38
rs1044523027 Chr.6: 115942368-115942368 on Build GRCh38
rs987554434 Chr.6: 115942342-115942342 on Build GRCh38
rs868320055 Chr.6: 115942485-115942485 on Build GRCh38
rs746440640 Chr.6: 115942392-115942392 on Build GRCh38
rs779278986 Chr.6: 115942373-115942373 on Build GRCh38
rs758361274 Chr.6: 115942400-115942402 on Build GRCh38
rs761052080 Chr.6: 115942422-115942422 on Build GRCh38
rs1003526937 Chr.6: 115942454-115942454 on Build GRCh38
rs1016687478 Chr.6: 115942491-115942491 on Build GRCh38
rs372823762 Chr.6: 115942468-115942468 on Build GRCh38
rs762906692 Chr.6: 115942482-115942482 on Build GRCh38
rs374931408 Chr.6: 115942395-115942395 on Build GRCh38
rs779092947 Chr.6: 115942503-115942503 on Build GRCh38
rs972651315 Chr.6: 115942512-115942512 on Build GRCh38
rs766247435 Chr.6: 115942483-115942483 on Build GRCh38
rs906176902 Chr.6: 115942399-115942399 on Build GRCh38
rs151160060 Chr.6: 115942535-115942535 on Build GRCh38
rs892044220 Chr.6: 115942463-115942463 on Build GRCh38
rs528046505 Chr.6: 115942444-115942444 on Build GRCh38
rs771106459 Chr.6: 115942370-115942370 on Build GRCh38
rs150528750 Chr.6: 115942490-115942490 on Build GRCh38
rs780554612 Chr.6: 115942530-115942530 on Build GRCh38
rs943510905 Chr.6: 115942347-115942347 on Build GRCh38
rs775887093 Chr.6: 115942419-115942419 on Build GRCh38
rs148599364 Chr.6: 115942495-115942495 on Build GRCh38
rs751339995 Chr.6: 115942484-115942484 on Build GRCh38
rs948823171 Chr.6: 115942366-115942366 on Build GRCh38
rs776243313 Chr.6: 115942540-115942540 on Build GRCh38
rs745969108 Chr.6: 115942505-115942505 on Build GRCh38
rs1003646414 Chr.6: 115942402-115942402 on Build GRCh38
rs761309616 Chr.6: 115942462-115942462 on Build GRCh38
rs147297157 Chr.6: 115942345-115942345 on Build GRCh38
rs12208531 Chr.6: 115942361-115942361 on Build GRCh38
rs371033823 Chr.6: 115942515-115942515 on Build GRCh38
rs113701747 Chr.6: 115942349-115942349 on Build GRCh38
rs199509116 Chr.6: 115942502-115942502 on Build GRCh38

View Full Product Details