Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 111515390-111515651 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf88
Entrez Gene ID
399949
Gene Name
chromosome 11 open reading frame 88
Gene Aliases
-
UniGene
Hs.411600
Gene Chromosome Location
Chr.11: 111514785-111537031 on Build GRCh38
Transcripts
NM_001100388.1 NM_207430.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6614148 c.260C>T p.A87V Chr.11: 111515544-111515544 on Build GRCh38
5837778 c.247T>G p.S83A Chr.11: 111515531-111515531 on Build GRCh38
2111539 c.248C>A p.S83Y Chr.11: 111515532-111515532 on Build GRCh38
4574040 c.260C>A p.A87E Chr.11: 111515544-111515544 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs780566009 Chr.11: 111515475-111515475 on Build GRCh38
rs372457530 Chr.11: 111515551-111515551 on Build GRCh38
rs1037030152 Chr.11: 111515439-111515439 on Build GRCh38
rs200124081 Chr.11: 111515570-111515570 on Build GRCh38
rs777714206 Chr.11: 111515508-111515508 on Build GRCh38
rs770821103 Chr.11: 111515514-111515514 on Build GRCh38
rs757579803 Chr.11: 111515465-111515465 on Build GRCh38
rs754685464 Chr.11: 111515467-111515467 on Build GRCh38
rs79930393 Chr.11: 111515510-111515510 on Build GRCh38
rs760039520 Chr.11: 111515471-111515471 on Build GRCh38
rs777191218 Chr.11: 111515599-111515599 on Build GRCh38
rs775797465 Chr.11: 111515553-111515554 on Build GRCh38
rs755614071 Chr.11: 111515595-111515595 on Build GRCh38
rs548171107 Chr.11: 111515491-111515491 on Build GRCh38
rs1017930935 Chr.11: 111515461-111515461 on Build GRCh38
rs767135790 Chr.11: 111515572-111515572 on Build GRCh38
rs529187598 Chr.11: 111515544-111515544 on Build GRCh38
rs372648264 Chr.11: 111515493-111515493 on Build GRCh38
rs560507006 Chr.11: 111515608-111515608 on Build GRCh38
rs769197034 Chr.11: 111515504-111515504 on Build GRCh38
rs368855991 Chr.11: 111515549-111515549 on Build GRCh38
rs969537059 Chr.11: 111515606-111515606 on Build GRCh38
rs200237513 Chr.11: 111515534-111515534 on Build GRCh38
rs374060793 Chr.11: 111515514-111515515 on Build GRCh38
rs551189239 Chr.11: 111515444-111515444 on Build GRCh38
rs779291081 Chr.11: 111515466-111515466 on Build GRCh38
rs775579194 Chr.11: 111515545-111515545 on Build GRCh38
rs12274967 Chr.11: 111515456-111515456 on Build GRCh38
rs752319642 Chr.11: 111515587-111515587 on Build GRCh38
rs765569734 Chr.11: 111515562-111515562 on Build GRCh38
rs549022678 Chr.11: 111515611-111515611 on Build GRCh38
rs750851481 Chr.11: 111515569-111515569 on Build GRCh38
rs762354787 Chr.11: 111515560-111515560 on Build GRCh38
rs12294424 Chr.11: 111515413-111515413 on Build GRCh38
rs527401399 Chr.11: 111515558-111515558 on Build GRCh38
rs138170508 Chr.11: 111515418-111515418 on Build GRCh38
rs754324137 Chr.11: 111515462-111515462 on Build GRCh38

View Full Product Details