Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 33264289-33264536 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAG1
Entrez Gene ID
573
Gene Name
BCL2 associated athanogene 1
Gene Aliases
BAG-1, HAP, RAP46
UniGene
Hs.377484
Gene Chromosome Location
Chr.9: 33252471-33264761 on Build GRCh38
Transcripts
NM_001172415.1 NM_004323.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6568676 c.241_252del12 p.T81_E84delTRSE Chr.9: 33264423-33264434 on Build GRCh38
4007132 c.38C>T p.S13L Chr.9: 33264292-33264292 on Build GRCh38
5812201 c.205A>T p.R69W Chr.9: 33264470-33264470 on Build GRCh38
6609557 c.165G>C p.G55G Chr.9: 33264510-33264510 on Build GRCh38
1108302 c.220A>G p.K74E Chr.9: 33264455-33264455 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748486254 Chr.9: 33264339-33264356 on Build GRCh38
rs765789829 Chr.9: 33264468-33264468 on Build GRCh38
rs745315959 Chr.9: 33264315-33264315 on Build GRCh38
rs540292399 Chr.9: 33264426-33264426 on Build GRCh38
rs780849402 Chr.9: 33264311-33264311 on Build GRCh38
rs747215409 Chr.9: 33264346-33264346 on Build GRCh38
rs772291483 Chr.9: 33264340-33264340 on Build GRCh38
rs758959032 Chr.9: 33264428-33264428 on Build GRCh38
rs764755467 Chr.9: 33264451-33264453 on Build GRCh38
rs755667250 Chr.9: 33264316-33264316 on Build GRCh38
rs759205636 Chr.9: 33264429-33264443 on Build GRCh38
rs368034599 Chr.9: 33264329-33264329 on Build GRCh38
rs754484490 Chr.9: 33264476-33264476 on Build GRCh38
rs773800857 Chr.9: 33264394-33264395 on Build GRCh38
rs565124425 Chr.9: 33264439-33264439 on Build GRCh38
rs764057728 Chr.9: 33264446-33264446 on Build GRCh38
rs774395261 Chr.9: 33264511-33264511 on Build GRCh38
rs768178220 Chr.9: 33264420-33264420 on Build GRCh38
rs769742000 Chr.9: 33264435-33264435 on Build GRCh38
rs560602319 Chr.9: 33264430-33264430 on Build GRCh38
rs574732669 Chr.9: 33264351-33264351 on Build GRCh38
rs199993128 Chr.9: 33264436-33264436 on Build GRCh38
rs62640045 Chr.9: 33264427-33264427 on Build GRCh38
rs879445518 Chr.9: 33264419-33264419 on Build GRCh38
rs796664636 Chr.9: 33264405-33264407 on Build GRCh38
rs767454301 Chr.9: 33264462-33264462 on Build GRCh38
rs779446080 Chr.9: 33264337-33264354 on Build GRCh38
rs762998621 Chr.9: 33264368-33264368 on Build GRCh38
rs770471036 Chr.9: 33264403-33264404 on Build GRCh38
rs62640046 Chr.9: 33264440-33264440 on Build GRCh38
rs375997019 Chr.9: 33264317-33264317 on Build GRCh38
rs750115998 Chr.9: 33264401-33264401 on Build GRCh38
rs750014766 Chr.9: 33264464-33264464 on Build GRCh38
rs767225121 Chr.9: 33264399-33264399 on Build GRCh38
rs773612275 Chr.9: 33264341-33264341 on Build GRCh38
rs145805568 Chr.9: 33264377-33264377 on Build GRCh38
rs373037713 Chr.9: 33264486-33264486 on Build GRCh38
rs756202350 Chr.9: 33264453-33264453 on Build GRCh38
rs746065877 Chr.9: 33264487-33264487 on Build GRCh38
rs755836999 Chr.9: 33264466-33264466 on Build GRCh38
rs776188663 Chr.9: 33264427-33264427 on Build GRCh38
rs763968294 Chr.9: 33264389-33264389 on Build GRCh38
rs771205724 Chr.9: 33264347-33264347 on Build GRCh38
rs574287414 Chr.9: 33264395-33264412 on Build GRCh38
rs60992409 Chr.9: 33264402-33264404 on Build GRCh38
rs201904238 Chr.9: 33264483-33264483 on Build GRCh38
rs180733352 Chr.9: 33264433-33264433 on Build GRCh38
rs749352210 Chr.9: 33264505-33264505 on Build GRCh38
rs752081823 Chr.9: 33264485-33264485 on Build GRCh38
rs754270851 Chr.9: 33264412-33264412 on Build GRCh38
rs751467121 Chr.9: 33264393-33264393 on Build GRCh38
rs779242380 Chr.9: 33264323-33264323 on Build GRCh38
rs752492326 Chr.9: 33264478-33264479 on Build GRCh38
rs753450723 Chr.9: 33264470-33264470 on Build GRCh38
rs776843073 Chr.9: 33264348-33264348 on Build GRCh38
rs11551681 Chr.9: 33264480-33264480 on Build GRCh38
rs746131931 Chr.9: 33264434-33264434 on Build GRCh38
rs757031406 Chr.9: 33264309-33264309 on Build GRCh38
rs759475755 Chr.9: 33264364-33264364 on Build GRCh38
rs751237919 Chr.9: 33264305-33264305 on Build GRCh38
rs140687411 Chr.9: 33264432-33264432 on Build GRCh38
rs536080013 Chr.9: 33264494-33264494 on Build GRCh38
rs144426804 Chr.9: 33264447-33264447 on Build GRCh38
rs376241694 Chr.9: 33264490-33264490 on Build GRCh38
rs867387917 Chr.9: 33264383-33264383 on Build GRCh38
rs761885636 Chr.9: 33264516-33264516 on Build GRCh38
rs749323018 Chr.9: 33264495-33264495 on Build GRCh38
rs995828044 Chr.9: 33264336-33264336 on Build GRCh38
rs757122686 Chr.9: 33264396-33264396 on Build GRCh38
rs779653465 Chr.9: 33264423-33264423 on Build GRCh38
rs747307347 Chr.9: 33264431-33264431 on Build GRCh38
rs765379810 Chr.9: 33264367-33264367 on Build GRCh38
rs768947715 Chr.9: 33264334-33264351 on Build GRCh38

View Full Product Details