Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 3489050-3489293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASPA
Entrez Gene ID
443
Gene Name
aspartoacylase
Gene Aliases
ACY2, ASP
UniGene
Hs.171142
Gene Chromosome Location
Chr.17: 3474106-3502534 on Build GRCh38
Transcripts
NM_000049.2 NM_001128085.1 XM_017024661.1 NM_001321336.1 NM_001321337.1

Gene Symbol
SPATA22
Entrez Gene ID
84690
Gene Name
spermatogenesis associated 22
Gene Aliases
NYD-SP20, NYDSP20
UniGene
Hs.351068
Gene Chromosome Location
Chr.17: 3440011-3513852 on Build GRCh38
Transcripts
NM_000049.2 NM_001128085.1 XM_017024661.1 NM_001321336.1 NM_001321337.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6888406 c.553G>A p.G185R Chr.17: 3489261-3489261 on Build GRCh38
6888407 c.554G>A p.G185E Chr.17: 3489262-3489262 on Build GRCh38
5758340 c.580C>G p.Q194E Chr.17: 3489288-3489288 on Build GRCh38
5448971 c.552A>G p.Q184Q Chr.17: 3489260-3489260 on Build GRCh38
3362085 c.556G>C p.V186L Chr.17: 3489264-3489264 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772264936 Chr.17: 3489261-3489261 on Build GRCh38
rs747825306 Chr.17: 3489156-3489156 on Build GRCh38
rs73311007 Chr.17: 3489169-3489169 on Build GRCh38
rs753389782 Chr.17: 3489215-3489215 on Build GRCh38
rs1029713018 Chr.17: 3489083-3489083 on Build GRCh38
rs756844626 Chr.17: 3489221-3489221 on Build GRCh38
rs1012357349 Chr.17: 3489178-3489178 on Build GRCh38
rs1023365853 Chr.17: 3489200-3489200 on Build GRCh38
rs375663977 Chr.17: 3489212-3489212 on Build GRCh38
rs745553536 Chr.17: 3489244-3489244 on Build GRCh38
rs778385612 Chr.17: 3489233-3489233 on Build GRCh38
rs750698855 Chr.17: 3489193-3489193 on Build GRCh38
rs536855227 Chr.17: 3489128-3489128 on Build GRCh38
rs1014551540 Chr.17: 3489247-3489247 on Build GRCh38
rs747438350 Chr.17: 3489265-3489265 on Build GRCh38
rs754689698 Chr.17: 3489198-3489198 on Build GRCh38
rs573646986 Chr.17: 3489168-3489168 on Build GRCh38
rs902481026 Chr.17: 3489138-3489138 on Build GRCh38
rs780468376 Chr.17: 3489262-3489262 on Build GRCh38
rs575664981 Chr.17: 3489105-3489105 on Build GRCh38
rs765354678 Chr.17: 3489188-3489188 on Build GRCh38
rs567140525 Chr.17: 3489152-3489152 on Build GRCh38
rs368144957 Chr.17: 3489201-3489201 on Build GRCh38
rs786204572 Chr.17: 3489249-3489249 on Build GRCh38
rs534647503 Chr.17: 3489158-3489158 on Build GRCh38
rs956754065 Chr.17: 3489097-3489097 on Build GRCh38
rs372085439 Chr.17: 3489202-3489202 on Build GRCh38

View Full Product Details