Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 58481694-58481963 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALPK2
Entrez Gene ID
115701
Gene Name
alpha kinase 2
Gene Aliases
HAK
UniGene
Hs.628152
Gene Chromosome Location
Chr.18: 58481247-58629192 on Build GRCh38
Transcripts
NM_052947.3 XM_011525801.2 XM_011525802.2 XM_017025538.1 XM_017025539.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
709261 c.4498A>T p.K1500* Chr.18: 58481921-58481921 on Build GRCh38
3371087 c.4516A>G p.I1506V Chr.18: 58481903-58481903 on Build GRCh38
1600052 c.4592C>A p.T1531N Chr.18: 58481827-58481827 on Build GRCh38
4503391 c.6395C>T p.S2132F Chr.18: 58481941-58481941 on Build GRCh38
6598778 c.6451C>A p.Q2151K Chr.18: 58481885-58481885 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61743671 Chr.18: 58481877-58481877 on Build GRCh38
rs7240666 Chr.18: 58481867-58481867 on Build GRCh38
rs143495285 Chr.18: 58481873-58481873 on Build GRCh38
rs914902329 Chr.18: 58481725-58481725 on Build GRCh38
rs750221423 Chr.18: 58481793-58481793 on Build GRCh38
rs201143538 Chr.18: 58481869-58481869 on Build GRCh38
rs536578168 Chr.18: 58481776-58481776 on Build GRCh38
rs557166287 Chr.18: 58481735-58481735 on Build GRCh38
rs752722370 Chr.18: 58481815-58481815 on Build GRCh38
rs546545381 Chr.18: 58481901-58481901 on Build GRCh38
rs750802597 Chr.18: 58481893-58481893 on Build GRCh38
rs940634045 Chr.18: 58481822-58481822 on Build GRCh38
rs759859454 Chr.18: 58481794-58481794 on Build GRCh38
rs774726690 Chr.18: 58481801-58481801 on Build GRCh38
rs749006100 Chr.18: 58481907-58481907 on Build GRCh38
rs571728845 Chr.18: 58481731-58481731 on Build GRCh38
rs3744867 Chr.18: 58481816-58481816 on Build GRCh38
rs138028987 Chr.18: 58481868-58481868 on Build GRCh38
rs572521431 Chr.18: 58481778-58481778 on Build GRCh38
rs767456166 Chr.18: 58481782-58481782 on Build GRCh38
rs546323829 Chr.18: 58481722-58481722 on Build GRCh38
rs371027763 Chr.18: 58481850-58481850 on Build GRCh38
rs761890043 Chr.18: 58481859-58481859 on Build GRCh38
rs969098128 Chr.18: 58481900-58481900 on Build GRCh38
rs148768721 Chr.18: 58481777-58481777 on Build GRCh38
rs950152212 Chr.18: 58481751-58481751 on Build GRCh38
rs35196844 Chr.18: 58481908-58481908 on Build GRCh38
rs202130652 Chr.18: 58481797-58481797 on Build GRCh38
rs368630136 Chr.18: 58481771-58481771 on Build GRCh38
rs59688801 Chr.18: 58481730-58481730 on Build GRCh38
rs772139303 Chr.18: 58481941-58481941 on Build GRCh38
rs751594533 Chr.18: 58481879-58481879 on Build GRCh38
rs375278224 Chr.18: 58481925-58481925 on Build GRCh38
rs532674203 Chr.18: 58481883-58481883 on Build GRCh38
rs761834474 Chr.18: 58481781-58481781 on Build GRCh38
rs747621408 Chr.18: 58481926-58481926 on Build GRCh38
rs996558411 Chr.18: 58481839-58481839 on Build GRCh38
rs530988474 Chr.18: 58481853-58481853 on Build GRCh38
rs772084467 Chr.18: 58481862-58481862 on Build GRCh38
rs769392146 Chr.18: 58481775-58481775 on Build GRCh38
rs550777054 Chr.18: 58481902-58481902 on Build GRCh38
rs779696708 Chr.18: 58481849-58481849 on Build GRCh38
rs372893643 Chr.18: 58481872-58481873 on Build GRCh38
rs779173243 Chr.18: 58481867-58481878 on Build GRCh38
rs149246242 Chr.18: 58481838-58481838 on Build GRCh38
rs771211292 Chr.18: 58481750-58481750 on Build GRCh38
rs138414829 Chr.18: 58481821-58481821 on Build GRCh38
rs752361224 Chr.18: 58481727-58481727 on Build GRCh38
rs773170762 Chr.18: 58481864-58481864 on Build GRCh38
rs543965909 Chr.18: 58481837-58481837 on Build GRCh38
rs767948648 Chr.18: 58481891-58481891 on Build GRCh38
rs538456739 Chr.18: 58481733-58481733 on Build GRCh38

View Full Product Details