Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 130693727-130693980 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC2
Entrez Gene ID
23404
Gene Name
exosome component 2
Gene Aliases
RRP4, Rrp4p, hRrp4p, p7
UniGene
Hs.654643
Gene Chromosome Location
Chr.9: 130693760-130704894 on Build GRCh38
Transcripts
NM_001282708.1 NM_001282709.1 NM_014285.6 NR_104230.1 XM_005272176.2 XM_006717023.2 XM_017014558.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4564000 c.9G>A p.M3I Chr.9: 130693800-130693800 on Build GRCh38
3721691 c.79G>C p.V27L Chr.9: 130693870-130693870 on Build GRCh38
1624632 c.24A>G p.P8P Chr.9: 130693815-130693815 on Build GRCh38
6657292 c.111A>G p.T37T Chr.9: 130693902-130693902 on Build GRCh38
4655443 c.117C>T p.F39F Chr.9: 130693908-130693908 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs970828188 Chr.9: 130693913-130693913 on Build GRCh38
rs762495185 Chr.9: 130693875-130693875 on Build GRCh38
rs759215090 Chr.9: 130693814-130693814 on Build GRCh38
rs762465859 Chr.9: 130693955-130693955 on Build GRCh38
rs748719679 Chr.9: 130693918-130693918 on Build GRCh38
rs565430068 Chr.9: 130693759-130693759 on Build GRCh38
rs762130666 Chr.9: 130693854-130693854 on Build GRCh38
rs758369646 Chr.9: 130693763-130693763 on Build GRCh38
rs771126752 Chr.9: 130693781-130693781 on Build GRCh38
rs766037616 Chr.9: 130693756-130693756 on Build GRCh38
rs749407451 Chr.9: 130693895-130693895 on Build GRCh38
rs200632112 Chr.9: 130693957-130693957 on Build GRCh38
rs146844600 Chr.9: 130693827-130693827 on Build GRCh38
rs780124515 Chr.9: 130693840-130693840 on Build GRCh38
rs758468229 Chr.9: 130693821-130693821 on Build GRCh38
rs769887590 Chr.9: 130693798-130693798 on Build GRCh38
rs915699181 Chr.9: 130693867-130693867 on Build GRCh38
rs754504282 Chr.9: 130693758-130693758 on Build GRCh38
rs367579263 Chr.9: 130693836-130693836 on Build GRCh38
rs576534270 Chr.9: 130693953-130693953 on Build GRCh38
rs780803058 Chr.9: 130693828-130693828 on Build GRCh38
rs145906216 Chr.9: 130693857-130693857 on Build GRCh38
rs374673172 Chr.9: 130693750-130693750 on Build GRCh38
rs749352427 Chr.9: 130693848-130693848 on Build GRCh38
rs751352040 Chr.9: 130693825-130693825 on Build GRCh38
rs199697937 Chr.9: 130693897-130693897 on Build GRCh38
rs934304177 Chr.9: 130693959-130693959 on Build GRCh38
rs763460421 Chr.9: 130693804-130693804 on Build GRCh38
rs376591547 Chr.9: 130693803-130693803 on Build GRCh38
rs752188322 Chr.9: 130693819-130693819 on Build GRCh38
rs963425075 Chr.9: 130693851-130693851 on Build GRCh38
rs200825653 Chr.9: 130693755-130693755 on Build GRCh38
rs781446909 Chr.9: 130693786-130693786 on Build GRCh38
rs769145406 Chr.9: 130693805-130693805 on Build GRCh38
rs757105392 Chr.9: 130693883-130693883 on Build GRCh38
rs765985454 Chr.9: 130693812-130693812 on Build GRCh38
rs773669995 Chr.9: 130693856-130693856 on Build GRCh38
rs778154366 Chr.9: 130693831-130693832 on Build GRCh38
rs981902740 Chr.9: 130693929-130693929 on Build GRCh38
rs781048882 Chr.9: 130693888-130693888 on Build GRCh38
rs777434826 Chr.9: 130693770-130693770 on Build GRCh38
rs554124783 Chr.9: 130693806-130693806 on Build GRCh38
rs773725040 Chr.9: 130693938-130693938 on Build GRCh38
rs755060021 Chr.9: 130693899-130693899 on Build GRCh38
rs756382553 Chr.9: 130693893-130693893 on Build GRCh38
rs371754404 Chr.9: 130693882-130693882 on Build GRCh38
rs764924683 Chr.9: 130693864-130693864 on Build GRCh38
rs748116494 Chr.9: 130693760-130693760 on Build GRCh38
rs148203698 Chr.9: 130693813-130693813 on Build GRCh38
rs769339881 Chr.9: 130693926-130693926 on Build GRCh38
rs747374184 Chr.9: 130693950-130693950 on Build GRCh38
rs780246147 Chr.9: 130693894-130693894 on Build GRCh38
rs752323145 Chr.9: 130693866-130693866 on Build GRCh38
rs1023725116 Chr.9: 130693896-130693896 on Build GRCh38
rs201877177 Chr.9: 130693749-130693749 on Build GRCh38
rs140638723 Chr.9: 130693849-130693849 on Build GRCh38
rs755941119 Chr.9: 130693797-130693797 on Build GRCh38
rs772506400 Chr.9: 130693855-130693855 on Build GRCh38
rs750165716 Chr.9: 130693891-130693891 on Build GRCh38
rs183616019 Chr.9: 130693809-130693809 on Build GRCh38
rs746122585 Chr.9: 130693830-130693830 on Build GRCh38
rs202001690 Chr.9: 130693823-130693823 on Build GRCh38
rs537467155 Chr.9: 130693880-130693880 on Build GRCh38
rs778808279 Chr.9: 130693917-130693917 on Build GRCh38
rs746806616 Chr.9: 130693776-130693776 on Build GRCh38
rs143849216 Chr.9: 130693815-130693815 on Build GRCh38
rs534366756 Chr.9: 130693782-130693782 on Build GRCh38

View Full Product Details