Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 38875057-38875316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJA9
Entrez Gene ID
81025
Gene Name
gap junction protein alpha 9
Gene Aliases
CX58, CX59, GJA10
UniGene
Hs.591506
Gene Chromosome Location
Chr.1: 38874067-38881626 on Build GRCh38
Transcripts
NM_030772.4 NR_037633.1 NR_037634.1 NR_037635.1 NR_037636.1 NR_037637.1 NR_135048.1

Gene Symbol
GJA9-MYCBP
Entrez Gene ID
100527950
Gene Name
GJA9-MYCBP readthrough
Gene Aliases
-
UniGene
Hs.591506
Gene Chromosome Location
Chr.1: 38862490-38881626 on Build GRCh38
Transcripts
NM_030772.4 NR_037633.1 NR_037634.1 NR_037635.1 NR_037636.1 NR_037637.1 NR_135048.1

Gene Symbol
LOC105378663
Entrez Gene ID
105378663
Gene Name
uncharacterized LOC105378663
Gene Aliases
-
UniGene
Hs.585419
Gene Chromosome Location
Chr.1: 38873401-38876226 on Build GRCh38
Transcripts
NM_030772.4 NR_037633.1 NR_037634.1 NR_037635.1 NR_037636.1 NR_037637.1 NR_135048.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
464573 c.1000T>G p.S334A Chr.1: 38875099-38875099 on Build GRCh38
909121 c.935A>G p.N312S Chr.1: 38875164-38875164 on Build GRCh38
5878604 c.918C>A p.F306L Chr.1: 38875181-38875181 on Build GRCh38
909120 c.954G>T p.E318D Chr.1: 38875145-38875145 on Build GRCh38
3751178 c.909T>C p.S303S Chr.1: 38875190-38875190 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762562353 Chr.1: 38875123-38875123 on Build GRCh38
rs147497931 Chr.1: 38875140-38875140 on Build GRCh38
rs945289810 Chr.1: 38875242-38875242 on Build GRCh38
rs756463977 Chr.1: 38875119-38875119 on Build GRCh38
rs753143985 Chr.1: 38875112-38875112 on Build GRCh38
rs767887017 Chr.1: 38875090-38875090 on Build GRCh38
rs749996613 Chr.1: 38875210-38875210 on Build GRCh38
rs1012007344 Chr.1: 38875228-38875228 on Build GRCh38
rs746934395 Chr.1: 38875229-38875229 on Build GRCh38
rs779569314 Chr.1: 38875224-38875224 on Build GRCh38
rs762134611 Chr.1: 38875215-38875215 on Build GRCh38
rs768572976 Chr.1: 38875237-38875237 on Build GRCh38
rs923492625 Chr.1: 38875126-38875126 on Build GRCh38
rs771181379 Chr.1: 38875255-38875255 on Build GRCh38
rs975003724 Chr.1: 38875120-38875120 on Build GRCh38
rs756583872 Chr.1: 38875200-38875200 on Build GRCh38
rs775262453 Chr.1: 38875157-38875157 on Build GRCh38
rs774585994 Chr.1: 38875263-38875263 on Build GRCh38
rs561351344 Chr.1: 38875202-38875202 on Build GRCh38
rs777022694 Chr.1: 38875173-38875173 on Build GRCh38
rs756898567 Chr.1: 38875132-38875132 on Build GRCh38
rs373832508 Chr.1: 38875137-38875137 on Build GRCh38
rs759502314 Chr.1: 38875191-38875191 on Build GRCh38
rs778148556 Chr.1: 38875122-38875122 on Build GRCh38
rs778689095 Chr.1: 38875209-38875209 on Build GRCh38
rs757879544 Chr.1: 38875221-38875221 on Build GRCh38
rs74804509 Chr.1: 38875167-38875167 on Build GRCh38
rs772098267 Chr.1: 38875156-38875156 on Build GRCh38
rs775570212 Chr.1: 38875270-38875270 on Build GRCh38
rs759752753 Chr.1: 38875269-38875269 on Build GRCh38
rs768359175 Chr.1: 38875159-38875159 on Build GRCh38
rs753164038 Chr.1: 38875194-38875194 on Build GRCh38
rs763166392 Chr.1: 38875171-38875171 on Build GRCh38
rs766574537 Chr.1: 38875182-38875182 on Build GRCh38
rs371970913 Chr.1: 38875208-38875208 on Build GRCh38
rs874243 Chr.1: 38875190-38875190 on Build GRCh38
rs187360719 Chr.1: 38875158-38875158 on Build GRCh38
rs748958511 Chr.1: 38875124-38875124 on Build GRCh38
rs142530971 Chr.1: 38875089-38875089 on Build GRCh38
rs530483739 Chr.1: 38875238-38875238 on Build GRCh38
rs368172759 Chr.1: 38875206-38875206 on Build GRCh38

View Full Product Details