Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129030702-129030957 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCC1
Entrez Gene ID
79825
Gene Name
EF-hand and coiled-coil domain containing 1
Gene Aliases
C3orf73, CCDC48
UniGene
Hs.134807
Gene Chromosome Location
Chr.3: 129001604-129040742 on Build GRCh38
Transcripts
NM_024768.2 XM_005247781.3 XM_011513161.2 XM_011513164.2 XM_017006941.1

Gene Symbol
KIAA1257
Entrez Gene ID
57501
Gene Name
KIAA1257
Gene Aliases
-
UniGene
Hs.518247
Gene Chromosome Location
Chr.3: 128910186-129032431 on Build GRCh38
Transcripts
NM_024768.2 XM_005247781.3 XM_011513161.2 XM_011513164.2 XM_017006941.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6392677 c.1127C>A p.A376D Chr.3: 129030849-129030849 on Build GRCh38
6842122 c.1135G>A p.E379K Chr.3: 129030857-129030857 on Build GRCh38
4521037 c.1090G>A p.G364R Chr.3: 129030812-129030812 on Build GRCh38
6836171 c.1122C>G p.S374R Chr.3: 129030844-129030844 on Build GRCh38
4631716 c.1007C>T p.P336L Chr.3: 129030729-129030729 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770992629 Chr.3: 129030875-129030875 on Build GRCh38
rs976196344 Chr.3: 129030725-129030725 on Build GRCh38
rs536035629 Chr.3: 129030899-129030899 on Build GRCh38
rs745592187 Chr.3: 129030894-129030894 on Build GRCh38
rs114454623 Chr.3: 129030730-129030730 on Build GRCh38
rs547264149 Chr.3: 129030827-129030827 on Build GRCh38
rs940994406 Chr.3: 129030771-129030771 on Build GRCh38
rs909452380 Chr.3: 129030760-129030760 on Build GRCh38
rs1002994926 Chr.3: 129030801-129030801 on Build GRCh38
rs1010803117 Chr.3: 129030721-129030721 on Build GRCh38
rs111306084 Chr.3: 129030908-129030908 on Build GRCh38
rs568707079 Chr.3: 129030853-129030853 on Build GRCh38
rs370403458 Chr.3: 129030807-129030807 on Build GRCh38
rs1035301295 Chr.3: 129030757-129030757 on Build GRCh38
rs960139984 Chr.3: 129030782-129030782 on Build GRCh38
rs1022899287 Chr.3: 129030834-129030834 on Build GRCh38
rs943114315 Chr.3: 129030936-129030936 on Build GRCh38
rs1036432619 Chr.3: 129030786-129030786 on Build GRCh38
rs906873344 Chr.3: 129030794-129030794 on Build GRCh38
rs768353590 Chr.3: 129030909-129030909 on Build GRCh38
rs557624250 Chr.3: 129030900-129030900 on Build GRCh38
rs1003379163 Chr.3: 129030733-129030733 on Build GRCh38
rs867779360 Chr.3: 129030738-129030738 on Build GRCh38
rs781194379 Chr.3: 129030880-129030880 on Build GRCh38
rs374365807 Chr.3: 129030889-129030889 on Build GRCh38
rs201241847 Chr.3: 129030729-129030729 on Build GRCh38
rs747147519 Chr.3: 129030855-129030855 on Build GRCh38
rs1049249510 Chr.3: 129030741-129030741 on Build GRCh38

View Full Product Details