Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 201833769-201834042 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK15
Entrez Gene ID
65061
Gene Name
cyclin dependent kinase 15
Gene Aliases
ALS2CR7, PFTAIRE2, PFTK2
UniGene
Hs.348711
Gene Chromosome Location
Chr.2: 201790324-201895550 on Build GRCh38
Transcripts
NM_001261435.1 NM_001261436.1 NM_139158.2 XM_005246781.3 XM_005246782.4 XM_011511650.2 XM_011511652.1 XM_011511654.2 XM_011511655.2 XM_011511656.2 XM_011511657.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5343965 c.454-1G>T p.? Chr.2: 201833847-201833847 on Build GRCh38
6625401 c.555A>G p.G185G Chr.2: 201833949-201833949 on Build GRCh38
1565151 c.465T>A p.F155L Chr.2: 201833859-201833859 on Build GRCh38
4505816 c.702C>T p.H234H Chr.2: 201833943-201833943 on Build GRCh38
5105845 c.577+10C>T p.? Chr.2: 201833981-201833981 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772862888 Chr.2: 201833855-201833855 on Build GRCh38
rs766268330 Chr.2: 201833816-201833816 on Build GRCh38
rs762881438 Chr.2: 201833986-201833986 on Build GRCh38
rs142697496 Chr.2: 201833898-201833898 on Build GRCh38
rs754427095 Chr.2: 201833837-201833837 on Build GRCh38
rs150762445 Chr.2: 201833871-201833871 on Build GRCh38
rs374936163 Chr.2: 201833925-201833925 on Build GRCh38
rs752302088 Chr.2: 201833934-201833934 on Build GRCh38
rs377138324 Chr.2: 201833833-201833833 on Build GRCh38
rs200976149 Chr.2: 201833979-201833979 on Build GRCh38
rs778256691 Chr.2: 201833932-201833932 on Build GRCh38
rs764608407 Chr.2: 201833880-201833880 on Build GRCh38
rs757637119 Chr.2: 201833832-201833832 on Build GRCh38
rs778541246 Chr.2: 201833840-201833840 on Build GRCh38
rs750825300 Chr.2: 201834003-201834003 on Build GRCh38
rs549866521 Chr.2: 201833847-201833847 on Build GRCh38
rs369534880 Chr.2: 201833995-201833995 on Build GRCh38
rs558726340 Chr.2: 201834008-201834008 on Build GRCh38
rs777726730 Chr.2: 201833936-201833936 on Build GRCh38
rs764896972 Chr.2: 201834010-201834010 on Build GRCh38
rs1030353473 Chr.2: 201833908-201833908 on Build GRCh38
rs775709227 Chr.2: 201833872-201833872 on Build GRCh38
rs746087465 Chr.2: 201833955-201833955 on Build GRCh38
rs752323951 Chr.2: 201834012-201834012 on Build GRCh38
rs753474054 Chr.2: 201833914-201833914 on Build GRCh38
rs752001844 Chr.2: 201833807-201833807 on Build GRCh38
rs372275183 Chr.2: 201833978-201833978 on Build GRCh38
rs370176322 Chr.2: 201833965-201833965 on Build GRCh38
rs114358447 Chr.2: 201833985-201833985 on Build GRCh38
rs773075862 Chr.2: 201833938-201833938 on Build GRCh38
rs781193371 Chr.2: 201833953-201833953 on Build GRCh38
rs145905632 Chr.2: 201833866-201833866 on Build GRCh38
rs765416594 Chr.2: 201833803-201833803 on Build GRCh38
rs368320214 Chr.2: 201833834-201833834 on Build GRCh38
rs1020309468 Chr.2: 201833827-201833827 on Build GRCh38
rs372228383 Chr.2: 201833810-201833810 on Build GRCh38
rs185846952 Chr.2: 201833804-201833804 on Build GRCh38
rs763343769 Chr.2: 201833879-201833879 on Build GRCh38
rs548488704 Chr.2: 201833886-201833886 on Build GRCh38
rs115873067 Chr.2: 201833870-201833870 on Build GRCh38
rs748029870 Chr.2: 201833846-201833846 on Build GRCh38
rs763045500 Chr.2: 201833981-201833981 on Build GRCh38
rs767862681 Chr.2: 201834002-201834002 on Build GRCh38
rs770796387 Chr.2: 201833950-201833950 on Build GRCh38
rs137880666 Chr.2: 201833869-201833869 on Build GRCh38
rs767881343 Chr.2: 201833899-201833899 on Build GRCh38

View Full Product Details