Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 19295901-19296144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND4C
Entrez Gene ID
55667
Gene Name
DENN domain containing 4C
Gene Aliases
C9orf55, C9orf55B, RAB10GEF, bA513M16.3
UniGene
Hs.249591
Gene Chromosome Location
Chr.9: 19230765-19374268 on Build GRCh38
Transcripts
NM_017925.6 NR_073201.3 XM_006716804.3 XM_006716805.3 XM_011517959.2 XM_017014917.1 XM_017014918.1 XM_017014919.1 XM_017014920.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
422404 c.217A>G p.T73A Chr.9: 19296131-19296131 on Build GRCh38
7082750 c.133C>T p.R45W Chr.9: 19296047-19296047 on Build GRCh38
4884225 c.129C>A p.Y43* Chr.9: 19296043-19296043 on Build GRCh38
6478434 c.191G>A p.R64K Chr.9: 19296105-19296105 on Build GRCh38
244009 c.179C>T p.T60M Chr.9: 19296093-19296093 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755264700 Chr.9: 19295987-19295987 on Build GRCh38
rs200586878 Chr.9: 19295968-19295968 on Build GRCh38
rs775510175 Chr.9: 19295963-19295963 on Build GRCh38
rs753264976 Chr.9: 19296005-19296005 on Build GRCh38
rs571142796 Chr.9: 19296010-19296010 on Build GRCh38
rs781436551 Chr.9: 19296023-19296023 on Build GRCh38
rs750723437 Chr.9: 19296096-19296096 on Build GRCh38
rs768302641 Chr.9: 19295965-19295968 on Build GRCh38
rs763501710 Chr.9: 19296088-19296088 on Build GRCh38
rs757295123 Chr.9: 19296094-19296094 on Build GRCh38
rs768690648 Chr.9: 19296047-19296047 on Build GRCh38
rs538781954 Chr.9: 19296058-19296058 on Build GRCh38
rs761343123 Chr.9: 19296056-19296056 on Build GRCh38
rs10757051 Chr.9: 19295943-19295943 on Build GRCh38
rs973382223 Chr.9: 19296040-19296040 on Build GRCh38
rs369894850 Chr.9: 19296067-19296067 on Build GRCh38
rs544315558 Chr.9: 19296002-19296002 on Build GRCh38
rs781119533 Chr.9: 19296095-19296095 on Build GRCh38
rs775420040 Chr.9: 19296033-19296033 on Build GRCh38
rs761423448 Chr.9: 19295983-19295983 on Build GRCh38
rs764614474 Chr.9: 19296090-19296090 on Build GRCh38
rs561253522 Chr.9: 19295933-19295933 on Build GRCh38
rs745655243 Chr.9: 19296027-19296027 on Build GRCh38
rs377299389 Chr.9: 19296064-19296064 on Build GRCh38
rs915323297 Chr.9: 19296084-19296084 on Build GRCh38
rs773754005 Chr.9: 19296053-19296053 on Build GRCh38
rs200037105 Chr.9: 19295998-19295998 on Build GRCh38
rs372328877 Chr.9: 19296012-19296012 on Build GRCh38
rs753032007 Chr.9: 19296079-19296079 on Build GRCh38
rs373799371 Chr.9: 19296071-19296071 on Build GRCh38
rs777872951 Chr.9: 19296011-19296011 on Build GRCh38
rs767152141 Chr.9: 19295984-19295984 on Build GRCh38
rs751736187 Chr.9: 19296093-19296093 on Build GRCh38
rs375027522 Chr.9: 19295958-19295959 on Build GRCh38
rs199516367 Chr.9: 19296110-19296110 on Build GRCh38
rs890651627 Chr.9: 19295950-19295950 on Build GRCh38
rs772868023 Chr.9: 19296063-19296063 on Build GRCh38
rs763765277 Chr.9: 19295975-19295975 on Build GRCh38
rs1028028131 Chr.9: 19296050-19296050 on Build GRCh38
rs749275954 Chr.9: 19296042-19296042 on Build GRCh38
rs757443750 Chr.9: 19296016-19296016 on Build GRCh38
rs1009001562 Chr.9: 19295974-19295974 on Build GRCh38

View Full Product Details