Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 209675802-209676043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
G0S2
Entrez Gene ID
50486
Gene Name
G0/G1 switch 2
Gene Aliases
-
UniGene
Hs.432132
Gene Chromosome Location
Chr.1: 209675325-209676390 on Build GRCh38
Transcripts
NM_015714.3 NR_134509.1 NR_134510.1

Gene Symbol
LOC101930114
Entrez Gene ID
101930114
Gene Name
uncharacterized LOC101930114
Gene Aliases
-
UniGene
Hs.663934
Gene Chromosome Location
Chr.1: 209661359-209742562 on Build GRCh38
Transcripts
NM_015714.3 NR_134509.1 NR_134510.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3966233 c.276C>T p.G92G Chr.1: 209675960-209675960 on Build GRCh38
5845335 c.236C>T p.A79V Chr.1: 209675920-209675920 on Build GRCh38
3740973 c.243G>C p.Q81H Chr.1: 209675927-209675927 on Build GRCh38
3671613 c.295C>A p.R99R Chr.1: 209675979-209675979 on Build GRCh38
292566 c.192G>A p.A64A Chr.1: 209675876-209675876 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140491567 Chr.1: 209675868-209675868 on Build GRCh38
rs766635565 Chr.1: 209675869-209675869 on Build GRCh38
rs749254329 Chr.1: 209676016-209676016 on Build GRCh38
rs759472154 Chr.1: 209675837-209675837 on Build GRCh38
rs753202670 Chr.1: 209675979-209675979 on Build GRCh38
rs150720329 Chr.1: 209675871-209675871 on Build GRCh38
rs142125926 Chr.1: 209675858-209675858 on Build GRCh38
rs765796722 Chr.1: 209675887-209675887 on Build GRCh38
rs775447110 Chr.1: 209675925-209675925 on Build GRCh38
rs967751564 Chr.1: 209676000-209676000 on Build GRCh38
rs73087483 Chr.1: 209676008-209676008 on Build GRCh38
rs899250673 Chr.1: 209675824-209675824 on Build GRCh38
rs775289692 Chr.1: 209675855-209675855 on Build GRCh38
rs766657848 Chr.1: 209675960-209675960 on Build GRCh38
rs982234446 Chr.1: 209675880-209675880 on Build GRCh38
rs890531673 Chr.1: 209676009-209676009 on Build GRCh38
rs778185736 Chr.1: 209675898-209675898 on Build GRCh38
rs780785373 Chr.1: 209675999-209675999 on Build GRCh38
rs764369651 Chr.1: 209675992-209675992 on Build GRCh38
rs142846097 Chr.1: 209675995-209675995 on Build GRCh38
rs750664680 Chr.1: 209675864-209675864 on Build GRCh38
rs376013217 Chr.1: 209675927-209675927 on Build GRCh38
rs756884533 Chr.1: 209675996-209675996 on Build GRCh38
rs781694971 Chr.1: 209675975-209675975 on Build GRCh38
rs770401244 Chr.1: 209675822-209675822 on Build GRCh38
rs145782263 Chr.1: 209675961-209675961 on Build GRCh38
rs745489332 Chr.1: 209676003-209676003 on Build GRCh38
rs765382333 Chr.1: 209675969-209675969 on Build GRCh38
rs143745964 Chr.1: 209675899-209675899 on Build GRCh38
rs753904643 Chr.1: 209675870-209675870 on Build GRCh38
rs780691591 Chr.1: 209675915-209675915 on Build GRCh38
rs140536045 Chr.1: 209675950-209675950 on Build GRCh38
rs769663352 Chr.1: 209675838-209675838 on Build GRCh38
rs753220688 Chr.1: 209675888-209675888 on Build GRCh38
rs929394301 Chr.1: 209675993-209675993 on Build GRCh38
rs762616506 Chr.1: 209675857-209675857 on Build GRCh38
rs758939231 Chr.1: 209675987-209675987 on Build GRCh38
rs774195177 Chr.1: 209675957-209675957 on Build GRCh38
rs199959013 Chr.1: 209675958-209675958 on Build GRCh38
rs776111464 Chr.1: 209675836-209675836 on Build GRCh38
rs780431905 Chr.1: 209675967-209675967 on Build GRCh38
rs779529696 Chr.1: 209676015-209676015 on Build GRCh38
rs758733987 Chr.1: 209675895-209675895 on Build GRCh38
rs977844273 Chr.1: 209676002-209676002 on Build GRCh38
rs368723492 Chr.1: 209675924-209675924 on Build GRCh38
rs756865282 Chr.1: 209675904-209675904 on Build GRCh38
rs745334619 Chr.1: 209675917-209675917 on Build GRCh38
rs759727145 Chr.1: 209675966-209675966 on Build GRCh38

View Full Product Details