Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 128019983-128020256 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD18
Entrez Gene ID
80167
Gene Name
abhydrolase domain containing 18
Gene Aliases
C4orf29
UniGene
Hs.143995
Gene Chromosome Location
Chr.4: 127965256-128039711 on Build GRCh38
Transcripts
NM_001319305.1 NM_001319306.1 NM_001319307.1 XM_005263238.4 XM_006714318.3 XM_006714320.3 XM_006714322.3 XM_011532272.2 XM_011532273.2 XM_011532274.2 XM_011532276.1 XM_011532278.2 XM_011532279.2 XM_017008631.1 XM_017008632.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4122201 c.614C>T p.A205V Chr.4: 128020084-128020084 on Build GRCh38
5589922 c.658C>G p.P220A Chr.4: 128020128-128020128 on Build GRCh38
5400474 c.674C>T p.S225F Chr.4: 128020144-128020144 on Build GRCh38
319094 c.679G>A p.A227T Chr.4: 128020149-128020149 on Build GRCh38
6313283 c.625G>T p.V209L Chr.4: 128020095-128020095 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747410756 Chr.4: 128020121-128020121 on Build GRCh38
rs375698416 Chr.4: 128020216-128020216 on Build GRCh38
rs745384841 Chr.4: 128020158-128020158 on Build GRCh38
rs762474035 Chr.4: 128020050-128020050 on Build GRCh38
rs781364186 Chr.4: 128020137-128020137 on Build GRCh38
rs757444653 Chr.4: 128020082-128020082 on Build GRCh38
rs770677812 Chr.4: 128020146-128020146 on Build GRCh38
rs1003221354 Chr.4: 128020229-128020229 on Build GRCh38
rs1015969705 Chr.4: 128020102-128020102 on Build GRCh38
rs781650594 Chr.4: 128020204-128020204 on Build GRCh38
rs768847777 Chr.4: 128020041-128020041 on Build GRCh38
rs747389849 Chr.4: 128020039-128020039 on Build GRCh38
rs775231143 Chr.4: 128020072-128020072 on Build GRCh38
rs755403120 Chr.4: 128020122-128020122 on Build GRCh38
rs868337589 Chr.4: 128020138-128020138 on Build GRCh38
rs1052073468 Chr.4: 128020231-128020231 on Build GRCh38
rs750590527 Chr.4: 128020181-128020181 on Build GRCh38
rs754392134 Chr.4: 128020155-128020155 on Build GRCh38
rs907846507 Chr.4: 128020157-128020157 on Build GRCh38
rs765436057 Chr.4: 128020180-128020180 on Build GRCh38
rs369691377 Chr.4: 128020060-128020060 on Build GRCh38
rs1044786778 Chr.4: 128020058-128020058 on Build GRCh38
rs746749687 Chr.4: 128020058-128020061 on Build GRCh38
rs375484020 Chr.4: 128020057-128020057 on Build GRCh38
rs772877780 Chr.4: 128020010-128020010 on Build GRCh38
rs762086816 Chr.4: 128020178-128020178 on Build GRCh38
rs766331669 Chr.4: 128020185-128020185 on Build GRCh38
rs202204733 Chr.4: 128020081-128020081 on Build GRCh38
rs758479961 Chr.4: 128020182-128020182 on Build GRCh38
rs765337908 Chr.4: 128020090-128020090 on Build GRCh38
rs201083117 Chr.4: 128020169-128020169 on Build GRCh38
rs774061812 Chr.4: 128020149-128020149 on Build GRCh38
rs763753545 Chr.4: 128020079-128020079 on Build GRCh38
rs974862964 Chr.4: 128020126-128020126 on Build GRCh38
rs772924492 Chr.4: 128020042-128020042 on Build GRCh38
rs537069265 Chr.4: 128020103-128020103 on Build GRCh38
rs770589859 Chr.4: 128020053-128020053 on Build GRCh38
rs760521555 Chr.4: 128020076-128020076 on Build GRCh38
rs749018442 Chr.4: 128020139-128020139 on Build GRCh38
rs767548321 Chr.4: 128020066-128020066 on Build GRCh38
rs761660121 Chr.4: 128020093-128020093 on Build GRCh38
rs1013919478 Chr.4: 128020007-128020007 on Build GRCh38
rs752130422 Chr.4: 128020195-128020195 on Build GRCh38
rs1045174541 Chr.4: 128020226-128020226 on Build GRCh38
rs1011221520 Chr.4: 128020088-128020088 on Build GRCh38
rs758965554 Chr.4: 128020094-128020094 on Build GRCh38
rs190456502 Chr.4: 128020056-128020056 on Build GRCh38
rs953838678 Chr.4: 128020226-128020226 on Build GRCh38
rs755420358 Chr.4: 128020197-128020197 on Build GRCh38
rs774107227 Chr.4: 128020167-128020167 on Build GRCh38
rs948224679 Chr.4: 128020225-128020226 on Build GRCh38
rs760477366 Chr.4: 128020168-128020168 on Build GRCh38

View Full Product Details