Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 3828215-3828488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLIS3
Entrez Gene ID
169792
Gene Name
GLIS family zinc finger 3
Gene Aliases
NDH, ZNF515
UniGene
Hs.162125
Gene Chromosome Location
Chr.9: 3824127-4310694 on Build GRCh38
Transcripts
NM_001042413.1 NM_152629.3 XM_005251386.4 XM_005251387.4 XM_005251388.4 XM_011517763.2 XM_011517764.2 XM_011517766.2 XM_011517767.2 XM_017014361.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4844492 c.2227G>A p.G743R Chr.9: 3828373-3828373 on Build GRCh38
6585812 c.2320G>T p.E774* Chr.9: 3828280-3828280 on Build GRCh38
1109106 c.2214C>A p.S738S Chr.9: 3828386-3828386 on Build GRCh38
5685516 c.2228G>A p.G743E Chr.9: 3828372-3828372 on Build GRCh38
3930065 c.2778C>G p.V926V Chr.9: 3828287-3828287 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766966152 Chr.9: 3828248-3828248 on Build GRCh38
rs767469455 Chr.9: 3828244-3828244 on Build GRCh38
rs377136249 Chr.9: 3828242-3828242 on Build GRCh38
rs756547082 Chr.9: 3828424-3828426 on Build GRCh38
rs150310830 Chr.9: 3828355-3828355 on Build GRCh38
rs561262659 Chr.9: 3828243-3828243 on Build GRCh38
rs774615723 Chr.9: 3828456-3828456 on Build GRCh38
rs780108058 Chr.9: 3828370-3828370 on Build GRCh38
rs369552895 Chr.9: 3828267-3828267 on Build GRCh38
rs377144389 Chr.9: 3828386-3828386 on Build GRCh38
rs776901417 Chr.9: 3828377-3828377 on Build GRCh38
rs754195885 Chr.9: 3828256-3828256 on Build GRCh38
rs759931240 Chr.9: 3828332-3828332 on Build GRCh38
rs76094493 Chr.9: 3828379-3828379 on Build GRCh38
rs772402767 Chr.9: 3828313-3828313 on Build GRCh38
rs1025472399 Chr.9: 3828398-3828398 on Build GRCh38
rs776073082 Chr.9: 3828450-3828450 on Build GRCh38
rs761079950 Chr.9: 3828247-3828247 on Build GRCh38
rs151140581 Chr.9: 3828319-3828319 on Build GRCh38
rs202213159 Chr.9: 3828457-3828457 on Build GRCh38
rs756799831 Chr.9: 3828352-3828352 on Build GRCh38
rs749999750 Chr.9: 3828391-3828391 on Build GRCh38
rs369030958 Chr.9: 3828344-3828344 on Build GRCh38
rs766979481 Chr.9: 3828390-3828390 on Build GRCh38
rs750979435 Chr.9: 3828346-3828346 on Build GRCh38
rs781124953 Chr.9: 3828295-3828295 on Build GRCh38
rs201132217 Chr.9: 3828387-3828387 on Build GRCh38
rs374480904 Chr.9: 3828451-3828451 on Build GRCh38
rs377640060 Chr.9: 3828432-3828432 on Build GRCh38
rs753240256 Chr.9: 3828312-3828319 on Build GRCh38
rs769039081 Chr.9: 3828455-3828455 on Build GRCh38
rs769595764 Chr.9: 3828306-3828306 on Build GRCh38
rs574827584 Chr.9: 3828261-3828261 on Build GRCh38
rs765676174 Chr.9: 3828334-3828334 on Build GRCh38
rs142480455 Chr.9: 3828296-3828296 on Build GRCh38
rs775310233 Chr.9: 3828309-3828309 on Build GRCh38
rs532596401 Chr.9: 3828340-3828340 on Build GRCh38
rs753509344 Chr.9: 3828405-3828405 on Build GRCh38
rs777119496 Chr.9: 3828281-3828281 on Build GRCh38
rs752421628 Chr.9: 3828275-3828275 on Build GRCh38
rs773378711 Chr.9: 3828246-3828246 on Build GRCh38
rs186788884 Chr.9: 3828461-3828461 on Build GRCh38
rs758121146 Chr.9: 3828280-3828280 on Build GRCh38
rs374173323 Chr.9: 3828269-3828269 on Build GRCh38
rs202168959 Chr.9: 3828384-3828384 on Build GRCh38
rs763191836 Chr.9: 3828452-3828452 on Build GRCh38
rs550506187 Chr.9: 3828458-3828458 on Build GRCh38
rs772126214 Chr.9: 3828360-3828360 on Build GRCh38
rs140101069 Chr.9: 3828342-3828342 on Build GRCh38
rs1002260300 Chr.9: 3828375-3828375 on Build GRCh38
rs749164251 Chr.9: 3828359-3828359 on Build GRCh38
rs746005781 Chr.9: 3828425-3828425 on Build GRCh38
rs979618943 Chr.9: 3828262-3828262 on Build GRCh38
rs377426046 Chr.9: 3828320-3828320 on Build GRCh38
rs57953443 Chr.9: 3828460-3828460 on Build GRCh38
rs780265238 Chr.9: 3828431-3828431 on Build GRCh38
rs779979488 Chr.9: 3828403-3828403 on Build GRCh38
rs765558063 Chr.9: 3828310-3828310 on Build GRCh38
rs1039901885 Chr.9: 3828369-3828369 on Build GRCh38
rs548436324 Chr.9: 3828378-3828378 on Build GRCh38
rs746600042 Chr.9: 3828282-3828282 on Build GRCh38
rs199920423 Chr.9: 3828287-3828287 on Build GRCh38
rs754832106 Chr.9: 3828408-3828408 on Build GRCh38
rs757132007 Chr.9: 3828285-3828285 on Build GRCh38
rs137946889 Chr.9: 3828356-3828356 on Build GRCh38
rs756029392 Chr.9: 3828401-3828401 on Build GRCh38
rs531799411 Chr.9: 3828371-3828371 on Build GRCh38
rs747234113 Chr.9: 3828421-3828421 on Build GRCh38
rs770369511 Chr.9: 3828449-3828449 on Build GRCh38
rs777308420 Chr.9: 3828329-3828329 on Build GRCh38
rs777809882 Chr.9: 3828412-3828412 on Build GRCh38
rs781055168 Chr.9: 3828353-3828353 on Build GRCh38
rs147357710 Chr.9: 3828312-3828312 on Build GRCh38
rs778060706 Chr.9: 3828368-3828368 on Build GRCh38
rs775707696 Chr.9: 3828383-3828383 on Build GRCh38
rs771158269 Chr.9: 3828373-3828373 on Build GRCh38
rs980534635 Chr.9: 3828434-3828434 on Build GRCh38
rs1031289713 Chr.9: 3828380-3828380 on Build GRCh38
rs773150809 Chr.9: 3828318-3828318 on Build GRCh38
rs771319961 Chr.9: 3828424-3828424 on Build GRCh38

View Full Product Details