Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 52028340-52028573 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG11
Entrez Gene ID
440138
Gene Name
ALG11, alpha-1,2-mannosyltransferase
Gene Aliases
CDG1P, GT8
UniGene
Hs.512963
Gene Chromosome Location
Chr.13: 52012387-52029648 on Build GRCh38
Transcripts
NM_001004127.2 NR_036571.2 NM_021645.5

Gene Symbol
UTP14C
Entrez Gene ID
9724
Gene Name
UTP14, small subunit processome component homolog C (S. cerevisiae)
Gene Aliases
2700066J21Rik, KIAA0266, UTP14B
UniGene
Hs.512963
Gene Chromosome Location
Chr.13: 52024691-52033600 on Build GRCh38
Transcripts
NM_001004127.2 NR_036571.2 NM_021645.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1607152 c.1451T>C p.L484P Chr.13: 52028562-52028562 on Build GRCh38
7174632 c.1389C>T p.I463I Chr.13: 52028500-52028500 on Build GRCh38
6598382 c.1257G>A p.S419S Chr.13: 52028368-52028368 on Build GRCh38
6001066 c.1290C>T p.H430H Chr.13: 52028401-52028401 on Build GRCh38
4215525 c.1256C>T p.S419L Chr.13: 52028367-52028367 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745722644 Chr.13: 52028472-52028472 on Build GRCh38
rs727503804 Chr.13: 52028420-52028420 on Build GRCh38
rs775344176 Chr.13: 52028489-52028489 on Build GRCh38
rs377356876 Chr.13: 52028402-52028402 on Build GRCh38
rs143100365 Chr.13: 52028388-52028388 on Build GRCh38
rs369694921 Chr.13: 52028380-52028380 on Build GRCh38
rs752810827 Chr.13: 52028457-52028457 on Build GRCh38
rs562562566 Chr.13: 52028442-52028442 on Build GRCh38
rs372199878 Chr.13: 52028454-52028454 on Build GRCh38
rs41292794 Chr.13: 52028518-52028518 on Build GRCh38
rs758597975 Chr.13: 52028458-52028458 on Build GRCh38
rs886039306 Chr.13: 52028513-52028513 on Build GRCh38
rs372405062 Chr.13: 52028387-52028387 on Build GRCh38
rs755514406 Chr.13: 52028440-52028440 on Build GRCh38
rs1053549004 Chr.13: 52028456-52028456 on Build GRCh38
rs746018813 Chr.13: 52028503-52028503 on Build GRCh38
rs201526759 Chr.13: 52028367-52028367 on Build GRCh38
rs371645290 Chr.13: 52028510-52028510 on Build GRCh38
rs373930425 Chr.13: 52028515-52028515 on Build GRCh38
rs199616330 Chr.13: 52028468-52028468 on Build GRCh38
rs763721498 Chr.13: 52028392-52028392 on Build GRCh38
rs762514154 Chr.13: 52028390-52028390 on Build GRCh38
rs951819111 Chr.13: 52028500-52028500 on Build GRCh38
rs757369277 Chr.13: 52028363-52028363 on Build GRCh38
rs769123692 Chr.13: 52028383-52028383 on Build GRCh38
rs779296066 Chr.13: 52028457-52028457 on Build GRCh38
rs768501691 Chr.13: 52028539-52028539 on Build GRCh38
rs769763630 Chr.13: 52028487-52028487 on Build GRCh38
rs759540582 Chr.13: 52028381-52028381 on Build GRCh38
rs751255090 Chr.13: 52028399-52028399 on Build GRCh38
rs368749306 Chr.13: 52028474-52028474 on Build GRCh38
rs773947350 Chr.13: 52028537-52028537 on Build GRCh38
rs766756419 Chr.13: 52028401-52028401 on Build GRCh38
rs757069433 Chr.13: 52028460-52028460 on Build GRCh38
rs770888492 Chr.13: 52028368-52028368 on Build GRCh38
rs199641594 Chr.13: 52028459-52028459 on Build GRCh38

View Full Product Details