Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35074858-35075131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCG
Entrez Gene ID
2189
Gene Name
Fanconi anemia complementation group G
Gene Aliases
FAG, XRCC9
UniGene
Hs.591084
Gene Chromosome Location
Chr.9: 35073838-35080016 on Build GRCh38
Transcripts
NM_004629.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4408697 c.1636+7A>G p.? Chr.9: 35074920-35074920 on Build GRCh38
1108584 c.1619G>A p.S540N Chr.9: 35074944-35074944 on Build GRCh38
6996493 c.1559G>A p.R520H Chr.9: 35075004-35075004 on Build GRCh38
4441849 c.1546G>A p.A516T Chr.9: 35075017-35075017 on Build GRCh38
1284879 c.1603C>G p.Q535E Chr.9: 35074960-35074960 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1000355215 Chr.9: 35074946-35074946 on Build GRCh38
rs758929136 Chr.9: 35074905-35074905 on Build GRCh38
rs778899700 Chr.9: 35074967-35074967 on Build GRCh38
rs753485145 Chr.9: 35074990-35074990 on Build GRCh38
rs772165907 Chr.9: 35074975-35074975 on Build GRCh38
rs1018397010 Chr.9: 35074965-35074965 on Build GRCh38
rs201422773 Chr.9: 35075018-35075018 on Build GRCh38
rs765540982 Chr.9: 35074999-35074999 on Build GRCh38
rs373151602 Chr.9: 35075044-35075044 on Build GRCh38
rs766086210 Chr.9: 35074927-35074927 on Build GRCh38
rs369348926 Chr.9: 35074907-35074907 on Build GRCh38
rs764379393 Chr.9: 35074883-35074883 on Build GRCh38
rs1003595944 Chr.9: 35075067-35075067 on Build GRCh38
rs45603633 Chr.9: 35074888-35074888 on Build GRCh38
rs17885240 Chr.9: 35075025-35075025 on Build GRCh38
rs768671803 Chr.9: 35074982-35074982 on Build GRCh38
rs763553947 Chr.9: 35075024-35075024 on Build GRCh38
rs760766384 Chr.9: 35074881-35074881 on Build GRCh38
rs776904008 Chr.9: 35074993-35074993 on Build GRCh38
rs754089024 Chr.9: 35074885-35074885 on Build GRCh38
rs968428980 Chr.9: 35074943-35074943 on Build GRCh38
rs370497978 Chr.9: 35075111-35075111 on Build GRCh38
rs763190093 Chr.9: 35074970-35074970 on Build GRCh38
rs755645109 Chr.9: 35075020-35075020 on Build GRCh38
rs747123733 Chr.9: 35074981-35074981 on Build GRCh38
rs376501845 Chr.9: 35074913-35074913 on Build GRCh38
rs587118 Chr.9: 35074920-35074920 on Build GRCh38
rs748364103 Chr.9: 35075071-35075071 on Build GRCh38
rs1012523223 Chr.9: 35075061-35075061 on Build GRCh38
rs748961871 Chr.9: 35074922-35074922 on Build GRCh38
rs752257189 Chr.9: 35074908-35074908 on Build GRCh38
rs1010004896 Chr.9: 35074914-35074914 on Build GRCh38
rs773509710 Chr.9: 35075093-35075093 on Build GRCh38
rs199833626 Chr.9: 35075017-35075017 on Build GRCh38
rs761993636 Chr.9: 35074996-35074996 on Build GRCh38
rs770011456 Chr.9: 35075082-35075082 on Build GRCh38
rs767443643 Chr.9: 35074972-35074974 on Build GRCh38
rs377131596 Chr.9: 35075080-35075080 on Build GRCh38
rs760738460 Chr.9: 35075080-35075080 on Build GRCh38
rs756990418 Chr.9: 35075026-35075026 on Build GRCh38
rs375978168 Chr.9: 35074904-35074904 on Build GRCh38
rs138395184 Chr.9: 35075043-35075043 on Build GRCh38
rs746983204 Chr.9: 35075051-35075051 on Build GRCh38
rs763292105 Chr.9: 35075098-35075098 on Build GRCh38
rs941422659 Chr.9: 35075075-35075075 on Build GRCh38
rs943779957 Chr.9: 35074986-35074986 on Build GRCh38
rs745748580 Chr.9: 35074971-35074971 on Build GRCh38
rs373411028 Chr.9: 35075042-35075042 on Build GRCh38
rs763387428 Chr.9: 35075005-35075005 on Build GRCh38
rs776600147 Chr.9: 35075068-35075068 on Build GRCh38
rs1045640267 Chr.9: 35075063-35075063 on Build GRCh38
rs768763154 Chr.9: 35075056-35075056 on Build GRCh38
rs906734281 Chr.9: 35075065-35075065 on Build GRCh38
rs369652577 Chr.9: 35075038-35075038 on Build GRCh38
rs761519737 Chr.9: 35074970-35074970 on Build GRCh38

View Full Product Details