Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 81061584-81061853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf46
Entrez Gene ID
123775
Gene Name
chromosome 16 open reading frame 46
Gene Aliases
-
UniGene
Hs.658684
Gene Chromosome Location
Chr.16: 81053497-81077267 on Build GRCh38
Transcripts
NM_001100873.1 NM_152337.2 XM_005255792.2 XM_011522848.2 XM_011522849.2 XM_017022916.1 XM_017022917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
974003 c.656C>A p.S219* Chr.16: 81061693-81061693 on Build GRCh38
4502795 c.622C>T p.L208F Chr.16: 81061727-81061727 on Build GRCh38
3421185 c.690G>T p.K230N Chr.16: 81061659-81061659 on Build GRCh38
6043007 c.606C>A p.P202P Chr.16: 81061743-81061743 on Build GRCh38
4406347 c.642G>A p.L214L Chr.16: 81061707-81061707 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs917869299 Chr.16: 81061642-81061642 on Build GRCh38
rs780292731 Chr.16: 81061675-81061675 on Build GRCh38
rs772149274 Chr.16: 81061769-81061769 on Build GRCh38
rs575038585 Chr.16: 81061712-81061712 on Build GRCh38
rs761730810 Chr.16: 81061814-81061814 on Build GRCh38
rs137925772 Chr.16: 81061823-81061823 on Build GRCh38
rs112523458 Chr.16: 81061614-81061614 on Build GRCh38
rs146937896 Chr.16: 81061773-81061773 on Build GRCh38
rs755158909 Chr.16: 81061762-81061762 on Build GRCh38
rs1053550861 Chr.16: 81061724-81061724 on Build GRCh38
rs754151927 Chr.16: 81061759-81061759 on Build GRCh38
rs752747522 Chr.16: 81061645-81061645 on Build GRCh38
rs575214348 Chr.16: 81061746-81061746 on Build GRCh38
rs748197489 Chr.16: 81061763-81061763 on Build GRCh38
rs138763234 Chr.16: 81061711-81061711 on Build GRCh38
rs529021063 Chr.16: 81061822-81061822 on Build GRCh38
rs931276281 Chr.16: 81061717-81061717 on Build GRCh38
rs746831628 Chr.16: 81061665-81061665 on Build GRCh38
rs201595004 Chr.16: 81061783-81061783 on Build GRCh38
rs764639607 Chr.16: 81061640-81061640 on Build GRCh38
rs371039198 Chr.16: 81061734-81061734 on Build GRCh38
rs780163711 Chr.16: 81061825-81061826 on Build GRCh38
rs758276184 Chr.16: 81061646-81061646 on Build GRCh38
rs375296318 Chr.16: 81061702-81061702 on Build GRCh38
rs542547879 Chr.16: 81061729-81061729 on Build GRCh38
rs150112740 Chr.16: 81061709-81061709 on Build GRCh38
rs778851312 Chr.16: 81061693-81061693 on Build GRCh38
rs759107267 Chr.16: 81061721-81061721 on Build GRCh38
rs748192210 Chr.16: 81061694-81061694 on Build GRCh38
rs804885 Chr.16: 81061758-81061758 on Build GRCh38
rs764170243 Chr.16: 81061651-81061651 on Build GRCh38
rs201993583 Chr.16: 81061753-81061753 on Build GRCh38
rs757009701 Chr.16: 81061666-81061666 on Build GRCh38
rs557650910 Chr.16: 81061738-81061738 on Build GRCh38
rs775171168 Chr.16: 81061796-81061796 on Build GRCh38
rs959254609 Chr.16: 81061608-81061608 on Build GRCh38
rs200487806 Chr.16: 81061710-81061710 on Build GRCh38
rs762412800 Chr.16: 81061797-81061797 on Build GRCh38
rs936305112 Chr.16: 81061716-81061716 on Build GRCh38
rs749487394 Chr.16: 81061690-81061690 on Build GRCh38
rs751553389 Chr.16: 81061811-81061811 on Build GRCh38
rs757089640 Chr.16: 81061756-81061756 on Build GRCh38
rs140831378 Chr.16: 81061626-81061626 on Build GRCh38
rs763569127 Chr.16: 81061806-81061806 on Build GRCh38
rs769358468 Chr.16: 81061784-81061784 on Build GRCh38
rs200288207 Chr.16: 81061816-81061816 on Build GRCh38
rs958938199 Chr.16: 81061791-81061791 on Build GRCh38
rs771489469 Chr.16: 81061775-81061775 on Build GRCh38
rs778474371 Chr.16: 81061770-81061770 on Build GRCh38
rs754567961 Chr.16: 81061618-81061618 on Build GRCh38
rs767371983 Chr.16: 81061815-81061815 on Build GRCh38
rs753076132 Chr.16: 81061742-81061742 on Build GRCh38
rs777140628 Chr.16: 81061782-81061782 on Build GRCh38
rs756510520 Chr.16: 81061744-81061749 on Build GRCh38

View Full Product Details