Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144291079-144291310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BOP1
Entrez Gene ID
23246
Gene Name
block of proliferation 1
Gene Aliases
-
UniGene
Hs.535901
Gene Chromosome Location
Chr.8: 144262046-144291438 on Build GRCh38
Transcripts
NM_015201.4

SNP Detail

SNP ID Chromosome Location
rs781817530 Chr.8: 144291253-144291253 on Build GRCh38
rs782619512 Chr.8: 144291245-144291245 on Build GRCh38
rs1011413076 Chr.8: 144291185-144291185 on Build GRCh38
rs978106017 Chr.8: 144291208-144291208 on Build GRCh38
rs782569588 Chr.8: 144291289-144291289 on Build GRCh38
rs907114777 Chr.8: 144291113-144291113 on Build GRCh38
rs782802564 Chr.8: 144291227-144291227 on Build GRCh38
rs782295654 Chr.8: 144291276-144291276 on Build GRCh38
rs782629928 Chr.8: 144291272-144291272 on Build GRCh38
rs782424807 Chr.8: 144291242-144291242 on Build GRCh38
rs781814254 Chr.8: 144291230-144291230 on Build GRCh38
rs782525259 Chr.8: 144291254-144291254 on Build GRCh38
rs1001344950 Chr.8: 144291123-144291123 on Build GRCh38
rs782537208 Chr.8: 144291283-144291283 on Build GRCh38
rs1024208933 Chr.8: 144291102-144291102 on Build GRCh38
rs558835221 Chr.8: 144291153-144291153 on Build GRCh38
rs967149762 Chr.8: 144291198-144291198 on Build GRCh38
rs1034163713 Chr.8: 144291131-144291131 on Build GRCh38
rs922874098 Chr.8: 144291217-144291217 on Build GRCh38
rs1022378394 Chr.8: 144291191-144291191 on Build GRCh38

View Full Product Details