Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 64925983-64926256 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKDD1A
Entrez Gene ID
348094
Gene Name
ankyrin repeat and death domain containing 1A
Gene Aliases
-
UniGene
Hs.207157
Gene Chromosome Location
Chr.15: 64911902-64958700 on Build GRCh38
Transcripts
NM_182703.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5383668 c.94G>A p.D32N Chr.15: 64926066-64926066 on Build GRCh38
5821010 c.382C>T p.L128L Chr.15: 64926081-64926081 on Build GRCh38
6226331 c.423G>A p.A141A Chr.15: 64926122-64926122 on Build GRCh38
2256128 c.392C>T p.A131V Chr.15: 64926091-64926091 on Build GRCh38
5887423 c.437A>G p.D146G Chr.15: 64926136-64926136 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546164052 Chr.15: 64926013-64926013 on Build GRCh38
rs74528237 Chr.15: 64926121-64926121 on Build GRCh38
rs748593551 Chr.15: 64926097-64926097 on Build GRCh38
rs1026081690 Chr.15: 64926006-64926006 on Build GRCh38
rs748925064 Chr.15: 64926082-64926082 on Build GRCh38
rs754916545 Chr.15: 64926063-64926063 on Build GRCh38
rs770234832 Chr.15: 64926194-64926194 on Build GRCh38
rs200619751 Chr.15: 64926042-64926042 on Build GRCh38
rs535983524 Chr.15: 64926204-64926204 on Build GRCh38
rs753354125 Chr.15: 64926215-64926215 on Build GRCh38
rs767366177 Chr.15: 64926124-64926124 on Build GRCh38
rs368473554 Chr.15: 64926200-64926200 on Build GRCh38
rs767562425 Chr.15: 64926064-64926064 on Build GRCh38
rs760052403 Chr.15: 64926049-64926049 on Build GRCh38
rs773231573 Chr.15: 64926107-64926107 on Build GRCh38
rs199876114 Chr.15: 64926095-64926095 on Build GRCh38
rs748681380 Chr.15: 64926190-64926190 on Build GRCh38
rs556174660 Chr.15: 64926149-64926149 on Build GRCh38
rs764090718 Chr.15: 64926212-64926212 on Build GRCh38
rs367970090 Chr.15: 64926202-64926202 on Build GRCh38
rs766596137 Chr.15: 64926120-64926120 on Build GRCh38
rs372737984 Chr.15: 64926089-64926089 on Build GRCh38
rs777353678 Chr.15: 64926079-64926079 on Build GRCh38
rs777103075 Chr.15: 64926074-64926074 on Build GRCh38
rs769865861 Chr.15: 64926100-64926100 on Build GRCh38
rs370741874 Chr.15: 64926075-64926075 on Build GRCh38
rs531442148 Chr.15: 64926026-64926026 on Build GRCh38
rs144050562 Chr.15: 64926162-64926162 on Build GRCh38
rs769184159 Chr.15: 64926032-64926032 on Build GRCh38
rs748879275 Chr.15: 64926188-64926188 on Build GRCh38
rs552484931 Chr.15: 64926161-64926161 on Build GRCh38
rs374819142 Chr.15: 64926209-64926209 on Build GRCh38
rs777760970 Chr.15: 64926198-64926198 on Build GRCh38
rs911989090 Chr.15: 64926155-64926155 on Build GRCh38
rs756680521 Chr.15: 64926027-64926028 on Build GRCh38
rs769107394 Chr.15: 64926091-64926091 on Build GRCh38
rs917527793 Chr.15: 64926157-64926157 on Build GRCh38
rs776732241 Chr.15: 64926040-64926040 on Build GRCh38
rs564353761 Chr.15: 64926021-64926021 on Build GRCh38
rs755899785 Chr.15: 64926020-64926020 on Build GRCh38
rs375747122 Chr.15: 64926130-64926130 on Build GRCh38
rs755348573 Chr.15: 64926173-64926173 on Build GRCh38
rs766114775 Chr.15: 64926061-64926061 on Build GRCh38
rs773434100 Chr.15: 64926053-64926053 on Build GRCh38
rs202207774 Chr.15: 64926218-64926218 on Build GRCh38
rs111873940 Chr.15: 64926027-64926027 on Build GRCh38
rs1046157061 Chr.15: 64926219-64926219 on Build GRCh38
rs748893030 Chr.15: 64926022-64926022 on Build GRCh38
rs61740365 Chr.15: 64926138-64926138 on Build GRCh38
rs538127589 Chr.15: 64926177-64926177 on Build GRCh38
rs768403279 Chr.15: 64926122-64926122 on Build GRCh38
rs763102108 Chr.15: 64926117-64926117 on Build GRCh38
rs114736780 Chr.15: 64926203-64926203 on Build GRCh38
rs376319037 Chr.15: 64926059-64926059 on Build GRCh38
rs761661222 Chr.15: 64926167-64926167 on Build GRCh38
rs370698584 Chr.15: 64926090-64926090 on Build GRCh38
rs980649492 Chr.15: 64926083-64926083 on Build GRCh38
rs528986828 Chr.15: 64926071-64926071 on Build GRCh38
rs149501643 Chr.15: 64926147-64926147 on Build GRCh38
rs150428985 Chr.15: 64926210-64926210 on Build GRCh38

View Full Product Details