Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 59182097-59182350 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTX4
Entrez Gene ID
23220
Gene Name
deltex E3 ubiquitin ligase 4
Gene Aliases
RNF155
UniGene
Hs.523696
Gene Chromosome Location
Chr.11: 59171430-59208587 on Build GRCh38
Transcripts
NM_001300727.1 NM_015177.1 XM_006718483.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4505293 c.371C>T p.P124L Chr.11: 59182216-59182216 on Build GRCh38
5767881 c.643C>T p.P215S Chr.11: 59182170-59182170 on Build GRCh38
2035184 c.757G>A p.A253T Chr.11: 59182284-59182284 on Build GRCh38
4679631 c.775C>T p.R259W Chr.11: 59182302-59182302 on Build GRCh38
5731930 c.421C>T p.R141* Chr.11: 59182266-59182266 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1041187006 Chr.11: 59182246-59182246 on Build GRCh38
rs764119701 Chr.11: 59182284-59182284 on Build GRCh38
rs911479640 Chr.11: 59182305-59182305 on Build GRCh38
rs377059900 Chr.11: 59182268-59182268 on Build GRCh38
rs778320668 Chr.11: 59182306-59182306 on Build GRCh38
rs199568679 Chr.11: 59182137-59182137 on Build GRCh38
rs201579194 Chr.11: 59182121-59182121 on Build GRCh38
rs760477316 Chr.11: 59182283-59182283 on Build GRCh38
rs61744443 Chr.11: 59182297-59182297 on Build GRCh38
rs201428116 Chr.11: 59182292-59182292 on Build GRCh38
rs571331933 Chr.11: 59182331-59182331 on Build GRCh38
rs368724184 Chr.11: 59182136-59182136 on Build GRCh38
rs771249081 Chr.11: 59182258-59182258 on Build GRCh38
rs766655274 Chr.11: 59182116-59182116 on Build GRCh38
rs1002418591 Chr.11: 59182214-59182214 on Build GRCh38
rs200628360 Chr.11: 59182219-59182219 on Build GRCh38
rs749633414 Chr.11: 59182257-59182257 on Build GRCh38
rs866434196 Chr.11: 59182191-59182191 on Build GRCh38
rs577423377 Chr.11: 59182192-59182192 on Build GRCh38
rs199998347 Chr.11: 59182193-59182193 on Build GRCh38
rs375750062 Chr.11: 59182325-59182325 on Build GRCh38
rs542889400 Chr.11: 59182267-59182267 on Build GRCh38
rs756823421 Chr.11: 59182179-59182179 on Build GRCh38
rs372682350 Chr.11: 59182303-59182303 on Build GRCh38
rs767762506 Chr.11: 59182133-59182133 on Build GRCh38
rs773400589 Chr.11: 59182250-59182250 on Build GRCh38
rs775722620 Chr.11: 59182269-59182269 on Build GRCh38
rs746545571 Chr.11: 59182302-59182302 on Build GRCh38
rs952725836 Chr.11: 59182162-59182162 on Build GRCh38
rs778600523 Chr.11: 59182180-59182180 on Build GRCh38
rs753764274 Chr.11: 59182286-59182286 on Build GRCh38
rs770348419 Chr.11: 59182238-59182238 on Build GRCh38
rs757057534 Chr.11: 59182289-59182289 on Build GRCh38
rs753685577 Chr.11: 59182160-59182160 on Build GRCh38
rs774857541 Chr.11: 59182327-59182327 on Build GRCh38
rs777292925 Chr.11: 59182237-59182237 on Build GRCh38
rs749815460 Chr.11: 59182312-59182312 on Build GRCh38
rs759664917 Chr.11: 59182123-59182123 on Build GRCh38
rs765078380 Chr.11: 59182291-59182291 on Build GRCh38
rs201955938 Chr.11: 59182266-59182266 on Build GRCh38
rs763734958 Chr.11: 59182144-59182144 on Build GRCh38

View Full Product Details