Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 103561840-103562103 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAG5
Entrez Gene ID
9529
Gene Name
BCL2 associated athanogene 5
Gene Aliases
BAG-5
UniGene
Hs.5443
Gene Chromosome Location
Chr.14: 103556544-103562814 on Build GRCh38
Transcripts
NM_001015048.2 NM_001015049.2 NM_004873.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4835524 c.81G>C p.W27C Chr.14: 103561927-103561927 on Build GRCh38
4432477 c.52G>C p.E18Q Chr.14: 103561956-103561956 on Build GRCh38
5428149 c.75A>G p.P25P Chr.14: 103561933-103561933 on Build GRCh38
5926881 c.25T>A p.L9M Chr.14: 103561983-103561983 on Build GRCh38
6609583 c.61A>G p.T21A Chr.14: 103561947-103561947 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550905580 Chr.14: 103561922-103561922 on Build GRCh38
rs781160077 Chr.14: 103561947-103561947 on Build GRCh38
rs775661661 Chr.14: 103561965-103561965 on Build GRCh38
rs778966498 Chr.14: 103561936-103561936 on Build GRCh38
rs115028071 Chr.14: 103561997-103561997 on Build GRCh38
rs964555022 Chr.14: 103562028-103562028 on Build GRCh38
rs7148456 Chr.14: 103561933-103561933 on Build GRCh38
rs749528440 Chr.14: 103562037-103562037 on Build GRCh38
rs768029686 Chr.14: 103561977-103561977 on Build GRCh38
rs375619271 Chr.14: 103562023-103562023 on Build GRCh38
rs749976469 Chr.14: 103561978-103561978 on Build GRCh38
rs199521433 Chr.14: 103562004-103562004 on Build GRCh38
rs769470164 Chr.14: 103561892-103561898 on Build GRCh38
rs764703608 Chr.14: 103561867-103561867 on Build GRCh38
rs780317327 Chr.14: 103562034-103562034 on Build GRCh38
rs774609803 Chr.14: 103562051-103562051 on Build GRCh38
rs772494977 Chr.14: 103562052-103562052 on Build GRCh38
rs397810530 Chr.14: 103561868-103561869 on Build GRCh38
rs748046116 Chr.14: 103561940-103561940 on Build GRCh38
rs952224455 Chr.14: 103562030-103562030 on Build GRCh38
rs371364108 Chr.14: 103561993-103561993 on Build GRCh38
rs774721334 Chr.14: 103561971-103561971 on Build GRCh38
rs1029904537 Chr.14: 103562076-103562076 on Build GRCh38
rs762066787 Chr.14: 103561891-103561892 on Build GRCh38
rs770053252 Chr.14: 103561959-103561959 on Build GRCh38
rs761447421 Chr.14: 103561882-103561882 on Build GRCh38
rs755863250 Chr.14: 103561894-103561894 on Build GRCh38
rs560447314 Chr.14: 103561887-103561887 on Build GRCh38
rs754600751 Chr.14: 103562005-103562005 on Build GRCh38
rs200468452 Chr.14: 103561909-103561909 on Build GRCh38
rs369258225 Chr.14: 103561875-103561875 on Build GRCh38
rs777206081 Chr.14: 103562064-103562064 on Build GRCh38
rs941122308 Chr.14: 103561883-103561883 on Build GRCh38
rs766950289 Chr.14: 103561884-103561884 on Build GRCh38
rs771975163 Chr.14: 103561946-103561946 on Build GRCh38
rs910605508 Chr.14: 103562077-103562077 on Build GRCh38
rs1044057175 Chr.14: 103561878-103561878 on Build GRCh38
rs377074898 Chr.14: 103561974-103561974 on Build GRCh38
rs1028337141 Chr.14: 103562011-103562011 on Build GRCh38
rs763284023 Chr.14: 103561866-103561866 on Build GRCh38
rs758288211 Chr.14: 103562019-103562019 on Build GRCh38
rs530834640 Chr.14: 103561913-103561913 on Build GRCh38
rs369702988 Chr.14: 103561957-103561957 on Build GRCh38
rs778573408 Chr.14: 103562013-103562013 on Build GRCh38
rs769862832 Chr.14: 103562026-103562026 on Build GRCh38
rs146624932 Chr.14: 103561931-103561931 on Build GRCh38
rs982418065 Chr.14: 103562084-103562084 on Build GRCh38
rs73361323 Chr.14: 103561966-103561966 on Build GRCh38
rs752492191 Chr.14: 103562016-103562016 on Build GRCh38
rs769132210 Chr.14: 103562003-103562003 on Build GRCh38
rs546904670 Chr.14: 103561874-103561874 on Build GRCh38
rs760319425 Chr.14: 103562056-103562056 on Build GRCh38
rs369774002 Chr.14: 103561869-103561869 on Build GRCh38
rs777662306 Chr.14: 103562021-103562021 on Build GRCh38
rs951629871 Chr.14: 103562083-103562083 on Build GRCh38
rs768892721 Chr.14: 103561970-103561970 on Build GRCh38
rs1054859160 Chr.14: 103561935-103561935 on Build GRCh38
rs768857299 Chr.14: 103562043-103562043 on Build GRCh38

View Full Product Details