Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 39177909-39178170 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNB1
Entrez Gene ID
782
Gene Name
calcium voltage-gated channel auxiliary subunit beta 1
Gene Aliases
CAB1, CACNLB1, CCHLB1
UniGene
Hs.635
Gene Chromosome Location
Chr.17: 39173456-39197703 on Build GRCh38
Transcripts
NM_000723.4 NM_199247.2 NM_199248.2 XM_005257645.2 XM_006722072.2 XM_017025024.1 XM_017025025.1 XM_017025026.1 XM_017025027.1 XM_017025028.1 XM_017025029.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4958549 c.1078G>A p.G360R Chr.17: 39178052-39178052 on Build GRCh38
6581937 c.1236C>A p.L412L Chr.17: 39178029-39178029 on Build GRCh38
5568183 c.1227C>T p.S409S Chr.17: 39178038-39178038 on Build GRCh38
5627814 c.1256C>T p.S419L Chr.17: 39178009-39178009 on Build GRCh38
5627813 c.1121C>T p.S374L Chr.17: 39178009-39178009 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs72556381 Chr.17: 39178014-39178014 on Build GRCh38
rs762338847 Chr.17: 39178128-39178128 on Build GRCh38
rs541746766 Chr.17: 39177959-39177959 on Build GRCh38
rs143114489 Chr.17: 39177985-39177985 on Build GRCh38
rs756203737 Chr.17: 39177955-39177955 on Build GRCh38
rs370951612 Chr.17: 39178088-39178088 on Build GRCh38
rs34356495 Chr.17: 39177965-39177966 on Build GRCh38
rs374604610 Chr.17: 39178111-39178111 on Build GRCh38
rs750613855 Chr.17: 39178059-39178059 on Build GRCh38
rs772947242 Chr.17: 39177986-39177986 on Build GRCh38
rs568685414 Chr.17: 39178148-39178148 on Build GRCh38
rs149931857 Chr.17: 39178094-39178094 on Build GRCh38
rs745492155 Chr.17: 39178117-39178117 on Build GRCh38
rs979798103 Chr.17: 39178130-39178130 on Build GRCh38
rs748055283 Chr.17: 39177960-39177960 on Build GRCh38
rs72556378 Chr.17: 39178110-39178110 on Build GRCh38
rs763467795 Chr.17: 39178015-39178015 on Build GRCh38
rs752008625 Chr.17: 39178027-39178027 on Build GRCh38
rs898584106 Chr.17: 39177961-39177961 on Build GRCh38
rs3744353 Chr.17: 39177990-39177990 on Build GRCh38
rs17857142 Chr.17: 39177988-39177988 on Build GRCh38
rs757687005 Chr.17: 39178034-39178034 on Build GRCh38
rs56041321 Chr.17: 39178125-39178125 on Build GRCh38
rs776261800 Chr.17: 39178008-39178008 on Build GRCh38
rs767906410 Chr.17: 39178053-39178053 on Build GRCh38
rs759027947 Chr.17: 39178009-39178009 on Build GRCh38
rs746007853 Chr.17: 39177942-39177942 on Build GRCh38
rs758414093 Chr.17: 39178107-39178107 on Build GRCh38
rs776280319 Chr.17: 39178116-39178116 on Build GRCh38
rs908332658 Chr.17: 39178019-39178019 on Build GRCh38
rs770611131 Chr.17: 39177991-39177991 on Build GRCh38
rs148965423 Chr.17: 39178147-39178147 on Build GRCh38
rs920626601 Chr.17: 39177969-39177969 on Build GRCh38
rs756155360 Chr.17: 39178078-39178078 on Build GRCh38
rs952094133 Chr.17: 39178151-39178151 on Build GRCh38
rs545970256 Chr.17: 39178016-39178016 on Build GRCh38
rs769209173 Chr.17: 39178121-39178121 on Build GRCh38

View Full Product Details